Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01301792_cn
          See other PRPF38B CNV Assays ›
          Gene Symbol
          PRPF38B
          Assay Reference Genome
          Location

          Chr.1:108698714 on build GRCh38
          Cytoband
          1p13.3
          Assay ID Hs01301792_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55119

          Gene Name:

          pre-mRNA processing factor 38B

          Gene Aliases:

          NET1

          Location:

          Chr.1:108692292-108701803 on Build GRCh38

          Assay Gene Location:

          Within Exon 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PRPF38B NM_018061.2 5 951 NP_060531.2
          NR_037185.1 6 1001
          XM_011541687.2 XP_011539989.1
          XM_017001596.1 5 456 XP_016857085.1
          XM_017001597.1 6 500 XP_016857086.1
          XM_017001598.1 6 2043 XP_016857087.1
          AK001192.1 5 745 BAA91546.1
          AL833950.1 6 973 CAH10702.1
          BC007757.2 5 925 AAH07757.1
          BC016296.1 5 921
          BC024275.1 5 925
          BC040127.1 5 938 AAH40127.1
          BC107801.1 5 678 AAI07802.1
          BC130346.1 5 697
          BC132963.1 5 697

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic non-DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          poly(A) RNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline