Search
Search

Entrez Gene ID: | 339524 |
Gene Name: | long intergenic non-protein coding RNA 1140 |
Gene Aliases: |
- |
Location: |
Chr.1:87129765-87169204 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01140 | NR_026985.1 | 4 | 803 | |
| NR_026986.1 | 3 | 603 | ||
| NR_026987.1 | 3 | 596 | ||
| NR_026988.1 | 3 | 499 | ||
| NR_026989.1 | ||||
| AL833352.1 | 4 | 827 | ||
| BI818944.1 | 3 | 409 | ||
| BM722117.1 | ||||
| DA398164.1 | ||||
| DA981741.1 | ||||
| DB100426.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23870 | Chr.1:87114725 - 87148575 on Build GRCh38 | Gain |
|