Search
Search

Entrez Gene ID: | 4170 |
Gene Name: | MCL1 apoptosis regulator, BCL2 family member |
Gene Aliases: |
BCL2L3, EAT, MCL1-ES, MCL1L, MCL1S, Mcl-1, TM, bcl2-L-3, mcl1/EAT |
Location: |
Chr.1:150574558-150579610 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCL1 | NM_001197320.2 | NP_001184249.1 | ||
| NM_021960.5 | NP_068779.1 | |||
| NM_182763.3 | NP_877495.1 | |||
| AA453505.1 | 1 | 305 | ||
| AF118124.1 | AAD13299.1 | |||
| AK294462.1 | BAG57694.1 | |||
| AK297217.1 | BAG59700.1 | |||
| AK300499.1 | BAG62213.1 | |||
| AK304775.1 | BAG65530.1 | |||
| AK312508.1 | BAG35409.1 | |||
| AK316267.1 | BAH14638.1 | |||
| BC017197.2 | AAH17197.1 | |||
| BC071897.1 | AAH71897.1 | |||
| BC107735.1 | AAI07736.1 | |||
| L08246.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006838 | Chr.1:150453465 - 150750307 on Build GRCh38 | Gain |
|
| nsv6322618 | Chr.1:150579001 - 150580100 on Build GRCh38 | Deletion |
|
| nsv4057833 | Chr.1:150439004 - 150716344 on Build GRCh38 | Duplication |
|
Set Membership: |
|