Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01439154_cn
          See other PNKD CNV Assays ›
          Gene Symbol
          PNKD
          Assay Reference Genome
          Location

          Chr.2:218274919 on build GRCh38
          Cytoband
          2q35
          Assay ID Hs01439154_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          25953

          Gene Name:

          paroxysmal nonkinesigenic dyskinesia

          Gene Aliases:

          BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2

          Location:

          Chr.2:218270392-218346793 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PNKD NM_015488.4 NP_056303.3
          XM_017003771.1 XP_016859260.1
          AB033010.1 BAA86498.1
          AK289867.1
          BC036457.2 AAH36457.1

          Target Gene Details

          Entrez Gene ID:

          64114

          Gene Name:

          transmembrane BAX inhibitor motif containing 1

          Gene Aliases:

          LFG3, MST100, MSTP100, PP1201, RECS1

          Location:

          Chr.2:218274192-218292577 on Build GRCh38

          Assay Gene Location:

          Within Exon 17
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TMBIM1 NM_001321427.1 13 2218 NP_001308356.1
          NM_001321428.1 12 1629 NP_001308357.1
          NM_001321429.1 13 1805 NP_001308358.1
          NM_001321430.1 13 1745 NP_001308359.1
          NM_001321432.1 12 1569 NP_001308361.1
          NM_001321433.1 13 1742 NP_001308362.1
          NM_001321435.1 13 1816 NP_001308364.1
          NM_001321436.1 13 1820 NP_001308365.1
          NM_001321438.1 11 1579 NP_001308367.1
          NM_022152.5 12 1644 NP_071435.2
          NR_135643.1 11 1402
          XM_011511627.1 12 1997 XP_011509929.1
          AB257295.1 12 1705
          AF193045.1 12 1558 AAG22473.1
          AK027755.1 10 1292 BAB55346.1
          AK075465.1 8 1119 BAC11636.1
          AK090618.1 13 2216
          AK093911.1 10 2156
          AK130380.1 12 1602
          AK130713.1 12 1580
          BC007980.2 12 1558 AAH07980.1
          BC013428.1 12 1622 AAH13428.1
          BC026348.1 12 1579 AAH26348.1
          BC026693.2 12 1590 AAH26693.1
          CA488388.1 2 757

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Intronic Non-exonic non-DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
          regulation of synaptic transmission, dopaminergic
          regulation of dopamine metabolic process
          negative regulation of neurotransmitter secretion
          neuromuscular process controlling posture
          negative regulation of catalytic activity
          negative regulation of establishment of protein localization to plasma membrane
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of Fas signaling pathway
          positive regulation of blood vessel remodeling
          hydroxyacylglutathione hydrolase activity
          protein binding
          metal ion binding
          death receptor binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline