Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01494717_cn
          See other ADCYAP1 CNV Assays ›
          Gene Symbol
          ADCYAP1
          Assay Reference Genome
          Location

          Chr.18:910165 on build GRCh38
          Cytoband
          18p11.32
          Assay ID Hs01494717_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          116

          Gene Name:

          adenylate cyclase activating polypeptide 1

          Gene Aliases:

          PACAP

          Location:

          Chr.18:904342-912172 on Build GRCh38

          Assay Gene Location:

          Within Exon 6
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ADCYAP1 NM_001099733.1 5 1180 NP_001093203.1
          NM_001117.4 4 1252 NP_001108.2
          XM_005258081.3 6 1579 XP_005258138.1
          S83513.1 4 1062 AAB21470.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv517252 Chr.18:902327 - 910634 on Build GRCh38 Loss ADCYAP1
          dgv5810n54 Chr.18:351004 - 2238246 on Build GRCh38 Gain TYMSOS CETN1 COLEC12 LOC388456 YES1 LOC105376854 CLUL1 ADCYAP1 TYMS LINC00470 ENOSF1
          nsv528169 Chr.18:551688 - 2240220 on Build GRCh38 Loss TYMSOS CETN1 LOC388456 YES1 CLUL1 ADCYAP1 TYMS LINC00470 ENOSF1
          dgv5811n54 Chr.18:841320 - 1798153 on Build GRCh38 Gain LOC388456 ADCYAP1 LINC00470
          nsv482935 Chr.18:10001 - 2900002 on Build GRCh38 Loss TYMSOS LOC101926997 COLEC12 LOC388456 YES1 LOC105376854 THOC1 CBX3P2 TYMS LINC00470 ENOSF1 METTL4 ROCK1P1 CETN1 USP14 EMILIN2 SMCHD1 CLUL1 ADCYAP1 MIR8078 NDC80
          nsv576228 Chr.18:69836 - 1506323 on Build GRCh38 Loss TYMSOS COLEC12 LOC388456 YES1 LOC105376854 THOC1 TYMS LINC00470 ENOSF1 ROCK1P1 CETN1 USP14 CLUL1 ADCYAP1 MIR8078

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          behavioral fear response
          histamine secretion
          negative regulation of acute inflammatory response to antigenic stimulus
          negative regulation of acute inflammatory response to non-antigenic stimulus
          activation of adenylate cyclase activity
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          cell-cell signaling
          female pregnancy
          regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of cell proliferation
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          negative regulation of muscle cell apoptotic process
          positive regulation of neuron projection development
          sensory perception of pain
          cAMP-mediated signaling
          pituitary gland development
          neuron projection development
          positive regulation of interleukin-6 production
          regulation of protein localization
          negative regulation of GTPase activity
          response to starvation
          negative regulation of potassium ion transport
          positive regulation of GTPase activity
          response to ethanol
          negative regulation of cell cycle
          positive regulation of protein kinase activity
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          ATP metabolic process
          positive regulation of synaptic transmission, glutamatergic
          regulation of postsynaptic membrane potential
          positive regulation of growth hormone secretion
          negative regulation of glial cell proliferation
          positive regulation of ERK1 and ERK2 cascade
          regulation of oligodendrocyte progenitor proliferation
          cellular response to glucocorticoid stimulus
          positive regulation of chemokine (C-C motif) ligand 5 production
          positive regulation of somatostatin secretion
          receptor signaling protein activity
          receptor binding
          neuropeptide hormone activity
          protein binding
          pituitary adenylate cyclase activating polypeptide activity
          pituitary adenylate cyclase-activating polypeptide receptor binding
          peptide hormone receptor binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline