Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01500679_cn
          See other MC1R CNV Assays ›
          Gene Symbol
          MC1R
          Assay Reference Genome
          Location

          Chr.16:89920109 on build GRCh38
          Cytoband
          16q24.3
          Assay ID Hs01500679_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4157

          Gene Name:

          melanocortin 1 receptor

          Gene Aliases:

          CMM5, MSH-R, SHEP2

          Location:

          Chr.16:89917879-89920977 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MC1R NM_002386.3 1 2231 NP_002377.4
          AK024232.1 1 2232
          AK054666.1 2 1202
          AK225902.1 1 1248
          BC007856.2 1 1230 AAH07856.1
          BC080622.1 1 1057 AAH80622.1
          X67594.1 1 1019 CAA47865.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2762221 Chr.16:89610380 - 90039318 on Build GRCh38 Gain+Loss CHMP1A LOC107984817 CENPBD1 GAS8 VPS9D1 DEF8 SPATA2L VPS9D1-AS1 FANCA DBNDD1 ZNF276 DPEP1 SPIRE2 CDK10 TUBB3 AFG3L1P SPATA33 LOC105371419 GAS8-AS1 MC1R TCF25
          nsv525671 Chr.16:89915950 - 89932386 on Build GRCh38 Loss MC1R TUBB3
          nsv573810 Chr.16:89857498 - 90037251 on Build GRCh38 Loss AFG3L1P CENPBD1 GAS8 DEF8 DBNDD1 GAS8-AS1 MC1R SPIRE2 TUBB3 TCF25
          nsv1070371 Chr.16:89919991 - 89925792 on Build GRCh38 Deletion MC1R TUBB3
          nsv428331 Chr.16:89216635 - 90185980 on Build GRCh38 Gain+Loss CHMP1A GAS8 DEF8 VPS9D1-AS1 URAHP LOC105371414 ZNF276 DPEP1 FAM157C SPIRE2 CDK10 TUBB3 SPG7 ANKRD11 LOC101927817 LOC107984817 PRDM7 CENPBD1 VPS9D1 SPATA2L FANCA SNORD68 DBNDD1 ZNF778 RPL13 AFG3L1P SPATA33 TUBB8P7 LOC105371419 GAS8-AS1 MC1R CPNE7 TCF25 LOC100287036
          nsv573799 Chr.16:89852186 - 90022379 on Build GRCh38 Gain AFG3L1P CENPBD1 GAS8 DEF8 DBNDD1 LOC105371419 MC1R SPIRE2 TUBB3 TCF25
          nsv509640 Chr.16:89899749 - 89969797 on Build GRCh38 Insertion CENPBD1 DEF8 MC1R TUBB3 TCF25
          nsv482951 Chr.16:88633593 - 90228345 on Build GRCh38 Loss GAS8 DEF8 VPS9D1-AS1 LOC107987238 FAM157C SPIRE2 CDK10 TUBB3 SPG7 CDT1 CTU2 IL17C RNF166 SLC22A31 LOC101927817 VPS9D1 FANCA PIEZO1 SNORD68 DBNDD1 ZNF778 LOC101927793 RPL13 AFG3L1P SPATA33 TUBB8P7 GAS8-AS1 MC1R SNAI3-AS1 TCF25 CHMP1A LOC100289580 URAHP LOC105371414 ZNF276 DPEP1 ANKRD11 ACSF3 MVD MIR4722 GALNS PABPN1L LOC105371409 LOC107984817 PRDM7 CENPBD1 SPATA2L CBFA2T3 LOC339059 SNAI3 LOC100129697 CDH15 LOC400558 TRAPPC2L APRT LINC00304 LOC105371419 CPNE7 LOC100287036 CYBA
          nsv1066499 Chr.16:89891957 - 90028891 on Build GRCh38 Loss AFG3L1P CENPBD1 GAS8 DEF8 DBNDD1 MC1R TUBB3 TCF25

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          multicellular organism development
          UV protection
          positive regulation of protein kinase A signaling
          sensory perception of pain
          positive regulation of cAMP biosynthetic process
          negative regulation of tumor necrosis factor production
          intracellular signal transduction
          melanin biosynthetic process
          pigmentation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of protein kinase B signaling
          UV-damage excision repair
          positive regulation of protein kinase C signaling
          melanocortin receptor activity
          melanocyte-stimulating hormone receptor activity
          protein binding
          G-protein coupled peptide receptor activity
          ubiquitin protein ligase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline