Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01504653_cn
          See other PFKL CNV Assays ›
          Gene Symbol
          PFKL
          Assay Reference Genome
          Location

          Chr.21:44304340 on build GRCh38
          Cytoband
          Assay ID Hs01504653_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5211

          Gene Name:

          phosphofructokinase, liver type

          Gene Aliases:

          ATP-PFK, PFK-B, PFK-L

          Location:

          Chr.21:44300053-44327373 on Build GRCh38

          Assay Gene Location:

          Within Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PFKL NM_001002021.3 2 204 NP_001002021.2
          NM_002626.6 NP_002617.3
          XM_005261135.4 XP_005261192.1
          XM_006724011.2 2 204 XP_006724074.1
          XM_017028369.2 2 204 XP_016883858.1
          XM_024452085.2 2 204 XP_024307853.1
          XM_047440822.1 2 204 XP_047296778.1
          XM_047440824.1 2 122 XP_047296780.1
          XM_047440825.1 2 204 XP_047296781.1
          AB209127.1 BAD92364.1
          AK091488.1
          AK303605.1 BAG64620.1
          AK308437.1
          BC006422.1 2 169 AAH06422.1
          BC007536.1 AAH07536.1
          BC008964.2 AAH08964.1
          BC009919.2 AAH09919.1
          CB997055.1
          HY111158.1
          X15573.1 CAA33597.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv953646 Chr.21:44279118 - 44523117 on Build GRCh38 Deletion LINC02575 LRRC3-DT LRRC3 TSPEAR-AS2 TSPEAR-AS1 AIRE PFKL TSPEAR CFAP410 TRPM2-AS TRPM2
          nsv1058359 Chr.21:44304172 - 44547662 on Build GRCh38 Gain LINC02575 LRRC3-DT LRRC3 TSPEAR-AS2 TSPEAR-AS1 PFKL TSPEAR CFAP410 TRPM2-AS KRTAP10-1 TRPM2
          esv2723638 Chr.21:44249174 - 44593709 on Build GRCh38 Deletion LINC02575 LRRC3-DT LRRC3 TSPEAR-AS2 TSPEAR-AS1 CFAP410 KRTAP10-6 KRTAP10-1 TRPM2 KRTAP10-2 DNMT3L KRTAP10-5 AIRE PFKL TSPEAR TRPM2-AS DNMT3L-AS1 KRTAP10-4 KRTAP10-3
          nsv509800 Chr.21:44284048 - 44471571 on Build GRCh38 Insertion LRRC3-DT LRRC3 AIRE PFKL CFAP410 TRPM2-AS TRPM2
          nsv6041254 Chr.21:44287180 - 44349861 on Build GRCh38 Deletion AIRE PFKL CFAP410
          nsv587779 Chr.21:44300477 - 44330463 on Build GRCh38 Loss PFKL CFAP410
          nsv587774 Chr.21:44283777 - 44306907 on Build GRCh38 Loss AIRE PFKL
          nsv587740 Chr.21:44209682 - 44429981 on Build GRCh38 Loss ICOSLG DNMT3L AIRE PFKL CFAP410 TRPM2-AS DNMT3L-AS1 TRPM2
          esv33817 Chr.21:43749531 - 44647625 on Build GRCh38 Loss LINC02575 GATD3 LRRC3-DT PDXK LRRC3 AATBC CFAP410 PWP2 KRTAP10-6 TRPM2 KRTAP10-2 KRTAP10-11 KRTAP10-8 KRTAP10-7 KRTAP10-10 PFKL TSPEAR TRPM2-AS CSTB ICOSLG TSPEAR-AS2 TSPEAR-AS1 TRAPPC10 KRTAP10-1 LINC01678 DNMT3L KRTAP10-5 RRP1 AIRE DNMT3L-AS1 LOC105372832 KRTAP10-9 AGPAT3 KRTAP10-4 KRTAP10-3
          nsv587780 Chr.21:44302559 - 44344514 on Build GRCh38 Loss PFKL CFAP410
          nsv4283103 Chr.21:43950580 - 44658488 on Build GRCh38 Duplication LINC02575 GATD3 LRRC3-DT KRTAP12-3 LRRC3 CFAP410 PWP2 KRTAP10-6 TRPM2 KRTAP10-2 KRTAP10-11 KRTAP10-8 KRTAP10-7 KRTAP10-10 PFKL TSPEAR KRTAP12-4 TRPM2-AS ICOSLG TSPEAR-AS2 TSPEAR-AS1 TRAPPC10 KRTAP10-1 LINC01678 DNMT3L KRTAP10-5 AIRE DNMT3L-AS1 LOC105372832 KRTAP10-9 AGPAT3 KRTAP10-4 KRTAP10-3

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose 6-phosphate metabolic process
          glycolytic process
          response to glucose
          fructose 1,6-bisphosphate metabolic process
          negative regulation of insulin secretion
          glycolytic process through fructose-6-phosphate
          canonical glycolysis
          nucleotide binding
          catalytic activity
          6-phosphofructokinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          kinase binding
          identical protein binding
          metal ion binding
          monosaccharide binding
          fructose binding
          fructose-6-phosphate binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline