Search
Search

Entrez Gene ID: | 10494 |
Gene Name: | serine/threonine kinase 25 |
Gene Aliases: |
SOK1, YSK1 |
Location: |
Chr.2:241494707-241509730 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK25 | NM_001271977.1 | 12 | 1788 | NP_001258906.1 |
| NM_001271978.1 | 11 | 1760 | NP_001258907.1 | |
| NM_001271979.1 | 11 | 1557 | NP_001258908.1 | |
| NM_001271980.1 | 11 | 1495 | NP_001258909.1 | |
| NM_001282305.1 | 11 | 1755 | NP_001269234.1 | |
| NM_001282306.1 | 12 | 1738 | NP_001269235.1 | |
| NM_001282307.1 | 11 | 1658 | NP_001269236.1 | |
| NM_001282308.1 | 11 | 1596 | NP_001269237.1 | |
| NM_006374.4 | 12 | 1726 | NP_006365.2 | |
| XM_011510494.2 | 12 | 1591 | XP_011508796.1 | |
| XM_011510495.2 | 11 | 1863 | XP_011508797.1 | |
| XM_017003168.1 | 11 | 1665 | XP_016858657.1 | |
| XM_017003169.1 | 11 | 1546 | XP_016858658.1 | |
| XM_017003170.1 | 11 | 1494 | XP_016858659.1 | |
| XM_017003171.1 | 10 | 1466 | XP_016858660.1 | |
| AK092161.1 | 11 | 1418 | ||
| AK291808.1 | 11 | 1384 | ||
| AK303719.1 | 11 | 1728 | ||
| AK315966.1 | ||||
| BC007852.2 | 12 | 1650 | AAH07852.1 | |
| BC015793.1 | 11 | 1706 | AAH15793.1 | |
| BC091505.1 | 12 | 1680 | AAH91505.1 | |
| CB250694.1 | 1 | 513 | ||
| CR936874.1 | 2 | 4452 | CAI59786.1 | |
| D63780.1 | 11 | 1549 | BAA20420.1 | |
| X99325.1 | 11 | 1561 | CAA67700.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759120 | Chr.2:241480540 - 241562243 on Build GRCh38 | Loss |
|
Set Membership: |
|