Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01773721_cn
          See other IPO9-AS1 CNV Assays ›
          Gene Symbol
          IPO9-AS1
          Assay Reference Genome
          Location

          Chr.1:201808572 on build GRCh38
          Cytoband
          1q32.1
          Assay ID Hs01773721_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          100873949

          Gene Name:

          IPO9 antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.1:201688256-201829559 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          IPO9-AS1 NR_046696.1

          Target Gene Details

          Entrez Gene ID:

          100302158

          Gene Name:

          microRNA 1231

          Gene Aliases:

          MIRN1231, hsa-mir-1231

          Location:

          Chr.1:201808611-201808702 on Build GRCh38

          Assay Gene Location:

          Overlaps - Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MIR1231 NR_031599.1

          Target Gene Details

          Entrez Gene ID:

          89796

          Gene Name:

          neuron navigator 1

          Gene Aliases:

          POMFIL3, STEERIN1, UNC53H1

          Location:

          Chr.1:201648322-201826974 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 20 - Intron 20
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NAV1 NM_001167738.1 NP_001161210.1
          NM_020443.4 NP_065176.3
          XM_006711609.2 XP_006711672.2
          XM_006711610.2 XP_006711673.2
          XM_006711611.1 XP_006711674.1
          XM_011510097.1 XP_011508399.1
          XM_011510098.1 XP_011508400.1
          XM_011510099.1 XP_011508401.1
          XM_011510100.1 XP_011508402.1
          XM_011510101.1 XP_011508403.1
          XM_011510102.1 XP_011508404.1
          XM_017002751.1 XP_016858240.1
          XM_017002752.1 XP_016858241.1
          AB032977.3 BAA86465.3
          AJ488101.1 CAD32470.1
          AK024265.1 BAB14865.1
          AY043013.1 AAL05591.1
          BC007523.1 AAH07523.1
          BC117683.1
          BX648460.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic non-DGV Variation

          Gene Ontology Categories:

          Function(s)

          microtubule bundle formation
          neuron migration

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline