Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01997970_cn
          See other OR2T10 CNV Assays ›
          Gene Symbol
          OR2T10
          Assay Reference Genome
          Location

          Chr.1:248593720 on build GRCh38
          Cytoband
          Assay ID Hs01997970_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          127069

          Gene Name:

          olfactory receptor family 2 subfamily T member 10

          Gene Aliases:

          OR1-64

          Location:

          Chr.1:232406-239620 on Build GRCh38

          Assay Gene Location:

          Within Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          OR2T10 NM_001004693.2 2 286 NP_001004693.1
          BC136937.1 1 80 AAI36938.1
          BC136940.1 1 80 AAI36941.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv630n100 Chr.1:248574134 - 248655170 on Build GRCh38 Gain+Loss OR2T10 OR2T35 OR2T27 OR2T34 OR2T11
          nsv4054120 Chr.1:248549154 - 248594102 on Build GRCh38 Deletion OR2T10 OR2T34 OR2T29
          nsv471517 Chr.1:248582659 - 248631802 on Build GRCh38 Loss OR2T10 OR2T11
          esv27363 Chr.1:248565805 - 248632217 on Build GRCh38 Loss OR2T10 OR2T34 OR2T11
          dgv625n100 Chr.1:248546303 - 248655170 on Build GRCh38 Gain OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2T29
          dgv58n172 Chr.1:248574700 - 248635099 on Build GRCh38 Deletion OR2T10 OR2T34 OR2T11
          dgv906n54 Chr.1:248330209 - 248694378 on Build GRCh38 Gain OR2T35 OR2T27 OR14C36 OR14I1 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2T3 OR2T6 OR2T5 OR2T34 OR2T1 OR2T11
          esv33052 Chr.1:248564627 - 248633381 on Build GRCh38 Gain+Loss OR2T10 OR2T34 OR2T11
          nsv433194 Chr.1:248581469 - 248652258 on Build GRCh38 Loss OR2T10 OR2T35 OR2T27 OR2T11
          dgv909n54 Chr.1:248525528 - 248631809 on Build GRCh38 Loss OR2T10 OR2T34 OR2T11 OR2G6 OR2T29
          nsv5438078 Chr.1:248564364 - 248654364 on Build GRCh38 Deletion OR2T10 OR2T35 OR2T27 OR2T34 OR2T11
          nsv6325373 Chr.1:248339419 - 248712414 on Build GRCh38 Duplication LYPD9P OR2T35 OR2T27 OR14C36 OR14I1 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2T3 OR2T6 OR2T5 OR2T34 OR2T1 OR2T11
          nsv4054201 Chr.1:248305738 - 248605228 on Build GRCh38 Deletion OR14C36 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2M7 OR2T3 OR2T6 OR2T5 OR2T34 OR2T1
          esv29940 Chr.1:248564598 - 248645127 on Build GRCh38 Loss OR2T10 OR2T35 OR2T34 OR2T11
          nsv4059817 Chr.1:248592373 - 248595182 on Build GRCh38 Deletion OR2T10
          dgv165e212 Chr.1:248520091 - 248670455 on Build GRCh38 Gain OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2G6 OR2T29
          nsv5376 Chr.1:248575503 - 248609055 on Build GRCh38 Insertion OR2T10
          dgv607n100 Chr.1:247895273 - 248661824 on Build GRCh38 Loss OR2T12 OR2T35 OR2T27 OR2L5 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 OR2L3 OR2W3 OR14C36 OR2M4 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 OR2L13 OR2T5 OR2T1
          dgv691n152 Chr.1:248549476 - 248649010 on Build GRCh38 Deletion OR2T10 OR2T35 OR2T34 OR2T11 OR2T29
          nsv1008002 Chr.1:248568732 - 248611786 on Build GRCh38 Loss OR2T10 OR2T34
          dgv637n100 Chr.1:248585881 - 248681796 on Build GRCh38 Loss OR2T10 OR2T35 OR2T27 OR2T11 OR14I1
          nsv442637 Chr.1:248585893 - 248633912 on Build GRCh38 Loss OR2T10 OR2T11
          nsv549641 Chr.1:248450347 - 248622144 on Build GRCh38 Gain OR2T10 OR2T2 OR2T3 OR2T5 OR2T34 OR2G6 OR2T29
          esv2763780 Chr.1:248520099 - 248650779 on Build GRCh38 Loss OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2G6 OR2T29
          dgv622n100 Chr.1:248520087 - 248670652 on Build GRCh38 Loss OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2G6 OR2T29
          dgv167e212 Chr.1:248589883 - 248631976 on Build GRCh38 Loss OR2T10 OR2T11
          nsv4069661 Chr.1:248567699 - 248634899 on Build GRCh38 Deletion OR2T10 OR2T34 OR2T11
          dgv627n100 Chr.1:248561921 - 248633900 on Build GRCh38 Loss OR2T10 OR2T34 OR2T11 OR2T29
          nsv1004790 Chr.1:248346193 - 248709345 on Build GRCh38 Gain LYPD9P OR2T35 OR2T27 OR14C36 OR14I1 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2T3 OR2T6 OR2T5 OR2T34 OR2T1 OR2T11
          dgv27n97 Chr.1:248561933 - 248662519 on Build GRCh38 Deletion OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2T29
          esv3589558 Chr.1:248592354 - 248595177 on Build GRCh38 Loss OR2T10
          nsv549603 Chr.1:247885685 - 248672148 on Build GRCh38 Loss OR2T12 OR2T35 OR2T27 OR2L5 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 OR2L3 OR2W3 OR14C36 OR2M4 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 OR2L13 OR2T5 OR2T1
          dgv201n166 Chr.1:247796651 - 248623796 on Build GRCh38 Duplication OR14A16 OR2T12 OR2L5 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 OR2L3 TRIM58 OR2W3 OR14C36 OR2M4 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 OR2L13 OR2T5 OR2T1 OR11L1
          nsv5983285 Chr.1:248560880 - 248602854 on Build GRCh38 Duplication OR2T10 OR2T34 OR2T29
          esv2750832 Chr.1:247897657 - 248873491 on Build GRCh38 Gain LYPD9P ZNF672 MIR3124 OR2T12 OR2T35 OR2T27 OR2L5 AHCYP8 OR2T29 OR2L1P PGBD2 OR2T2 OR2M7 OR2T3 OR2M1P OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 SH3BP5L OR2L3 OR14C36 OR2M4 OR14I1 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 ZNF692-DT OR2L13 OR2T5 OR2T1 LYPD8 ZNF692
          dgv132n145 Chr.1:248525274 - 248632837 on Build GRCh38 Deletion OR2T10 OR2T34 OR2T11 OR2G6 OR2T29
          dgv918n54 Chr.1:248586905 - 248638700 on Build GRCh38 Loss OR2T10 OR2T35 OR2T11
          nsv6319287 Chr.1:248461234 - 248695001 on Build GRCh38 Duplication OR2T10 OR2T3 OR2T35 OR2T27 OR2T5 OR2T34 OR2T11 OR14I1 OR2G6 OR2T29
          nsv515858 Chr.1:247620588 - 248705477 on Build GRCh38 Gain+Loss LYPD9P OR14A16 OR2T12 OR2T35 OR2T27 OR13G1 OR2L5 OR1C1 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR9H1 OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR14L1 OR2L8 OR2AJ1 OR2M2 OR2L3 OR6F1 TRIM58 OR2W3 OR14C36 OR2M4 OR14I1 OR2G6 OR2T7 OR2T10 LOC102724446 OR2T4 OR2AK2 OR2T6 OR14A2 OR2L13 OR2T5 OR14K1 OR2T1 OR11L1
          esv2760083 Chr.1:248442451 - 248655182 on Build GRCh38 Gain+Loss OR2T7 OR2T10 OR2T2 OR2T3 OR2T35 OR2T27 OR2T5 OR2T34 OR2T11 OR2G6 OR2T29
          dgv608n100 Chr.1:247907795 - 248631151 on Build GRCh38 Gain OR2T12 OR2L5 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 OR2L3 OR14C36 OR2M4 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 OR2L13 OR2T5 OR2T1
          nsv3169954 Chr.1:248550690 - 248650779 on Build GRCh38 Deletion OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2T29
          nsv428357 Chr.1:248336807 - 248714122 on Build GRCh38 Loss LYPD9P OR2T35 OR2T27 OR14C36 OR14I1 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2T3 OR2T6 OR2T5 OR2T34 OR2T1 OR2T11
          dgv617n100 Chr.1:248457862 - 248605673 on Build GRCh38 Gain OR2T10 OR2T3 OR2T5 OR2T34 OR2G6 OR2T29
          dgv29n111 Chr.1:248546369 - 248654081 on Build GRCh38 Deletion OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2T29
          esv2660478 Chr.1:248576044 - 248635091 on Build GRCh38 Deletion OR2T10 OR2T11
          nsv821623 Chr.1:248481643 - 248627107 on Build GRCh38 Loss OR2T10 OR2T5 OR2T34 OR2T11 OR2G6 OR2T29
          nsv1068651 Chr.1:248574798 - 248634999 on Build GRCh38 Deletion OR2T10 OR2T11
          esv3589559 Chr.1:248593440 - 248634296 on Build GRCh38 Loss OR2T10 OR2T11
          nsv1013273 Chr.1:247656673 - 248784560 on Build GRCh38 Gain LYPD9P OR14A16 OR2T12 OR2T35 OR2T27 OR13G1 OR2L5 AHCYP8 OR1C1 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR9H1 OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 OR2L3 OR6F1 TRIM58 OR2W3 OR14C36 OR2M4 OR14I1 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 OR14A2 OR2L13 OR2T5 OR14K1 OR2T1 OR11L1 LYPD8
          dgv133n145 Chr.1:248574916 - 248632837 on Build GRCh38 Deletion OR2T10 OR2T11
          dgv17e55 Chr.1:248079226 - 248654081 on Build GRCh38 Loss OR2T12 OR2T35 OR2M2 OR2T27 OR14C36 OR2M4 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2M7 OR2T3 OR2M1P OR2T6 OR2L13 OR2M5 OR2T5 OR2T34 OR2T1 OR2M3 OR2T11 OR2T33
          dgv116n67 Chr.1:248564398 - 248633377 on Build GRCh38 Loss OR2T10 OR2T34 OR2T11
          nsv469750 Chr.1:248538645 - 248714122 on Build GRCh38 Loss OR2T10 LYPD9P OR2T35 OR2T27 OR2T34 OR2T11 OR14I1 OR2T29
          nsv6139372 Chr.1:248370288 - 248775095 on Build GRCh38 Duplication LYPD9P OR2T35 OR2T27 OR14I1 AHCYP8 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T3 OR2T6 OR2T5 OR2T34 OR2T1 OR2T11 LYPD8
          dgv639n100 Chr.1:248593445 - 248655170 on Build GRCh38 Gain OR2T10 OR2T35 OR2T27 OR2T11
          nsv9191 Chr.1:248574482 - 248632893 on Build GRCh38 Gain+Loss OR2T10 OR2T34 OR2T11
          dgv916n54 Chr.1:248582901 - 248631809 on Build GRCh38 Loss OR2T10 OR2T11
          dgv911n54 Chr.1:248568548 - 248631809 on Build GRCh38 Loss OR2T10 OR2T34 OR2T11
          dgv134n145 Chr.1:248591107 - 248632837 on Build GRCh38 Deletion OR2T10 OR2T11
          nsv5874143 Chr.1:248549151 - 248594101 on Build GRCh38 Deletion OR2T10 OR2T34 OR2T29
          esv2421359 Chr.1:248576215 - 248633912 on Build GRCh38 Deletion OR2T10 OR2T11
          dgv166e212 Chr.1:248524920 - 248664922 on Build GRCh38 Loss OR2T10 OR2T35 OR2T27 OR2T34 OR2T11 OR2G6 OR2T29
          nsv5365 Chr.1:248552133 - 248635949 on Build GRCh38 Deletion OR2T10 OR2T34 OR2T11 OR2T29
          nsv5870440 Chr.1:248592368 - 248595181 on Build GRCh38 Deletion OR2T10
          esv2750835 Chr.1:248359378 - 248864040 on Build GRCh38 Gain LYPD9P ZNF672 MIR3124 OR2T35 OR2T27 SH3BP5L OR14I1 AHCYP8 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2T3 OR2T6 ZNF692-DT OR2T5 OR2T34 OR2T1 OR2T11 LYPD8 ZNF692
          nsv1160483 Chr.1:248475325 - 248650779 on Build GRCh38 Duplication OR2T10 OR2T35 OR2T27 OR2T5 OR2T34 OR2T11 OR2G6 OR2T29
          esv3891415 Chr.1:247876411 - 248795772 on Build GRCh38 Gain LYPD9P OR2T12 OR2T35 OR2T27 OR2L5 AHCYP8 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 OR2L3 TRIM58 OR2W3 OR14C36 OR2M4 OR14I1 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 OR2L13 OR2T5 OR2T1 LYPD8
          nsv827864 Chr.1:248590345 - 248632198 on Build GRCh38 Loss OR2T10 OR2T11
          dgv618n100 Chr.1:248475313 - 248633900 on Build GRCh38 Loss OR2T10 OR2T5 OR2T34 OR2T11 OR2G6 OR2T29
          nsv514048 Chr.1:248590684 - 248632156 on Build GRCh38 Loss OR2T10 OR2T11
          dgv584n223 Chr.1:247906731 - 248623795 on Build GRCh38 Duplication OR2T12 OR2L5 OR2T29 OR2L1P OR2T2 OR2M7 OR2T3 OR2M1P OR2T8 OR2M5 OR2T34 OR2M3 OR2L2 OR2T11 OR2T33 OR2L8 OR2AJ1 OR2M2 OR2L3 OR14C36 OR2M4 OR2G6 OR2T7 OR2T10 OR2T4 OR2AK2 OR2T6 OR2L13 OR2T5 OR2T1
          dgv20n17 Chr.1:248324336 - 248684068 on Build GRCh38 Loss OR2T35 OR2T27 OR14C36 OR14I1 OR2G6 OR2T29 OR2T7 OR2T10 OR2T2 OR2T4 OR2M7 OR2T3 OR2T6 OR2T5 OR2T34 OR2T1 OR2T11

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline