Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02079908_cn
          See other PKD1 CNV Assays ›
          Gene Symbol
          PKD1
          Assay Reference Genome
          Location

          Chr.16:2090516 on build GRCh38
          Cytoband
          Assay ID Hs02079908_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5310

          Gene Name:

          polycystin 1, transient receptor potential channel interacting

          Gene Aliases:

          PBP, PC1, Pc-1, TRPP1, eliosin

          Location:

          Chr.16:705067-760109 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 49 - Exon 50
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PKD1 NM_000296.4 NP_000287.4
          NM_001009944.3 NP_001009944.3
          XM_005255370.4 XP_005255427.1
          XM_011522528.4 XP_011520830.1
          XM_011522529.3 XP_011520831.1
          XM_011522537.2 XP_011520839.1
          XM_047434208.1 XP_047290164.1
          XM_047434209.1 XP_047290165.1
          XM_047434210.1 XP_047290166.1
          XM_047434211.1 XP_047290167.1
          XM_047434212.1 XP_047290168.1
          XM_047434213.1 XP_047290169.1
          AK309422.1
          L33243.1 AAC37576.1
          PQ628107.1
          U24497.1 AAC50128.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv4862n54 Chr.16:2088868 - 2095128 on Build GRCh38 Loss PKD1-AS1 MIR1225 TSC2 PKD1
          esv2422427 Chr.16:10001 - 2723164 on Build GRCh38 Duplication WDR24 LOC102723753 NPIPB5 CALB2 ATP6V0C LOC105371038 MRPS34 LOC105371129 BFAR CORO7-PAM16 MIR3677HG CDR2 PRSS27 ABCA17P MSRB1 MIR3670-3 MIR4717 SNHG9 IGFALS RNF151 DECR2 CENPN HMOX2 PDPK1 FAM234A MSLN SNTB2 MIR6511A1 CORO7 UTP4 LOC100287175 WDR90 CRAMP1 SMG1P2 MIR6859-4 MSLNL HBA1 PKD1P6-NPIPP1 TELO2 EME2 CCDC78 MIR6770-2 BMERB1 LINC02124 PKD1-AS1 STUB1 MIR6511A2 NPIPA1 RHBDL1 MIR940 SSTR5 NPW RPS2 CES5A NPIPB3 MIR6768 MAPK8IP3-AS1 GNPTG LOC100288162 IFT140 PRSS29P MIR6767 NDE1 TPSB2 FAHD1 ECI1 NHERF2 CLCN7 TSR3 LOC102723713 LOC131696449 RAB40C RRN3P3 MIR484 MPV17L C16orf46 POLR3K NTN3 LOC124907782 ABCA15P FBXL16 TPSG1 MIR3179-2 MIR6511A3 PARN CIAO3 LOC100505915 ZNF598 LUC7L CHTF18 MIR3670-1 CTRB2 BRICD5 NPIPB11 CACNA1H TMEM204 OTOAP1 KCTD5 MAPK8IP3 SLC7A5P2 WFIKKN1 SMG1P3 MIR3179-3 LOC124903619 PIGQ AXIN1 LINC00235 POLR3E SDR42E2 TNRC6A VWA3A PDZD9 METRN PTX4 PERCC1 LOC100190986 GFER PRSS30P MIR662 DNASE1L2 NPIPA8 OTOA NPIPA3 CRYM VASN SMG1P1 MIR3680-1 HS3ST6 NTHL1 BAIAP3 SOX8 IGSF6 MIR6770-1 LINC02890 PDIA2 PAM16 JMJD8 MIR3179-1 MIR5587 EEF2K ARHGAP17 UNKL-AS1 DNAJA3 RPL23AP5 MRPL28 MIR3180-2 MIR3680-2 ANKS4B GNG13 PKD1P2 SNHG19 ABCC1 NHLRC4 MIR3180-3 LOC101927814 MIR3677 MIR3180-1 CCDC154 SNX29P1 ECI1-AS1 CMC2 NDUFB10 ZP2 CEMP1 RAB26 MFSD13B MIR3180-4 LOC100134368 MARF1 CES1 RPUSD1 NME3 PKD1P1 MIR3178 MPV17L-BMERB1 CEROX1 FLJ42627 TPSD1 HBM MIR3180-5 MIR1972-1 RGS11 LOC102723692 HYDIN GLIS2-AS1 PRR35 RHOT2 METTL9 ZG16B SYNGR3 UBE2I HBA2 LOC728138 LOC105371046 NOMO3 LOC100506281 NPIPA7 ARHGDIG CASKIN1 LOC107987382 LOC107984876 RHBDF1 UNKL MIR1972-2 MLST8 METTL26 NPIPA5 LOC102724014 HAGH ABCC6P2 NPIPB4 TBL3 LOC124903621 MIR3177 NPIPA9 TBC1D24 ERVK13-1 RRN3 GCSH HBZ MIR3176 ABCA3 PDXDC1 MIR6506 SNORD60 MOSMO PRSS22 LOC107987386 TPSAB1 CDIP1 LOC105379545 SLC6A10PB TEDC2-AS1 TEDC2 CAPN15 SNORA10 LOC102723655 TSC2 PKD1 CTRB1 NME4 ANTKMT HAGHL ABCC6 STUB1-DT XYLT1 UBE2I-AS1 LMF1 E4F1 NMRAL1 LOC101929566 LOC102724052 RRN3P1 GLIS2 TRAF7 CEP20 AMDHD2 PKD1L2 PGAP6 LOC652276 CDR2-DT BCAR1 MIR3670-2 LINC00254 CMTR2 BCO1 PLA2G10 SNORA64 SNORA78 NOXO1 MIR4516 MEIOB C1QTNF8 SSTR5-AS1 NPIPA6 SMG1P6 PGP NPRL3 NOMO1 LMF1-AS1 UQCRC2 DDX11L10 LOC105371050 NUBP2 SLC5A11 CCNF ABCC6P1 UQCC4 SPSB3 RPL3L HBQ1 JPT2 SNRNP25 ATMIN NTAN1 MYH11 MPG RAB11FIP3 MIR6511B1 MIR1225 MCRIP2 MIR6511B2 WASIR2 RNPS1
          dgv4859n54 Chr.16:1971892 - 2264400 on Build GRCh38 Loss LOC131696449 PKD1P2 SNHG19 NOXO1 NPIPA7 MIR4516 MIR6770-2 SNORD60 CASKIN1 GFER ECI1-AS1 PKD1-AS1 DNASE1L2 NPIPA6 RAB26 PGP MIR6511A2 MLST8 MIR6511A3 TSC2 PKD1 ZNF598 NTHL1 PKD1P1 E4F1 NPW BRICD5 TBL3 MIR3180-5 TRAF7 MIR6511B1 MIR1225 ECI1 NHERF2 SYNGR3 RNPS1
          nsv952901 Chr.16:1888800 - 2135899 on Build GRCh38 Deletion LOC131696449 PKD1P2 SNORA64 NOMO3 SNORA78 NOXO1 NPIPA7 MIR4516 MIR6770-2 LINC02124 GFER NDUFB10 PKD1-AS1 NPIPA6 SNORA10 MIR3179-2 MIR6511A2 MIR6511A3 TSC2 PKD1 ZNF598 HS3ST6 NTHL1 MSRB1 PKD1P1 SNHG9 RNF151 NPW RPS2 TBL3 RPL3L MIR6511B1 MIR1225 NHERF2 MIR3180-2 MIR3670-2 SYNGR3
          nsv1160329 Chr.16:1961727 - 2128574 on Build GRCh38 Deletion LOC131696449 PKD1P2 SNORA64 SNORA78 NOXO1 NPIPA7 MIR6770-2 GFER NDUFB10 PKD1-AS1 NPIPA6 SNORA10 MIR6511A2 MIR6511A3 TSC2 PKD1 ZNF598 NTHL1 PKD1P1 SNHG9 RNF151 NPW RPS2 TBL3 MIR6511B1 MIR1225 NHERF2 MIR3180-2 MIR3670-2 SYNGR3
          dgv4861n54 Chr.16:2083900 - 2246850 on Build GRCh38 Loss E4F1 SNHG19 MIR4516 BRICD5 SNORD60 CASKIN1 MIR3180-5 ECI1-AS1 PKD1-AS1 DNASE1L2 TRAF7 MIR6511B1 MIR1225 RAB26 PGP ECI1 MLST8 TSC2 PKD1
          nsv3239719 Chr.16:2062481 - 2121886 on Build GRCh38 Insertion PKD1-AS1 MIR6511B1 MIR1225 TSC2 PKD1
          nsv571190 Chr.16:2055054 - 2200730 on Build GRCh38 Loss PKD1-AS1 TRAF7 SNHG19 MIR6511B1 MIR1225 MIR4516 RAB26 SNORD60 TSC2 CASKIN1 PKD1 MIR3180-5
          nsv1052260 Chr.16:2074546 - 2629655 on Build GRCh38 Gain ABCA3 SNHG19 ATP6V0C MIR4516 SNORD60 MIR3677 CASKIN1 ECI1-AS1 NTN3 PKD1-AS1 DNASE1L2 CEMP1 MIR3677HG TEDC2-AS1 TEDC2 RAB26 PGP MLST8 TSC2 PKD1 LOC105371050 ABCA17P MIR4717 MIR940 MIR3178 E4F1 CCNF BRICD5 PDPK1 MIR6768 MIR3180-5 TRAF7 MIR6767 AMDHD2 MIR6511B1 MIR1225 LOC652276 ECI1 TBC1D24 RNPS1
          nsv471381 Chr.16:2088710 - 2135898 on Build GRCh38 Gain PKD1-AS1 MIR6511B1 MIR1225 MIR4516 TSC2 PKD1

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          blood vessel development
          in utero embryonic development
          kidney development
          liver development
          embryonic placenta development
          protein export from nucleus
          calcium ion transport
          homophilic cell adhesion via plasma membrane adhesion molecules
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          cell surface receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          JAK-STAT cascade
          heart development
          anatomical structure morphogenesis
          organ morphogenesis
          regulation of gene expression
          regulation of cell cycle process
          Wnt signaling pathway
          spinal cord development
          neural tube development
          establishment of cell polarity
          regulation of cell adhesion
          response to fluid shear stress
          lymph vessel morphogenesis
          chordate embryonic development
          skin development
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          branching morphogenesis of an epithelial tube
          genitalia development
          detection of mechanical stimulus
          cartilage development
          protein heterotetramerization
          regulation of cell cycle
          regulation of mitotic spindle organization
          lung epithelium development
          placenta blood vessel development
          regulation of proteasomal protein catabolic process
          calcium ion transmembrane transport
          nitrogen cycle metabolic process
          mesonephric tubule development
          mesonephric duct development
          metanephric collecting duct development
          metanephric ascending thin limb development
          metanephric proximal tubule development
          metanephric distal tubule morphogenesis
          cell-cell adhesion
          mitocytosis
          regulation of G1/S transition of mitotic cell cycle
          cation channel activity
          calcium channel activity
          protein binding
          protein kinase binding
          protein domain specific binding
          carbohydrate binding
          Wnt-activated receptor activity
          ion channel binding
          transcription regulator inhibitor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline