Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02099650_cn
          See other IQSEC3 CNV Assays ›
          Gene Symbol
          IQSEC3
          Assay Reference Genome
          Location

          Chr.12:171332 on build GRCh38
          Cytoband
          12p13.33
          Assay ID Hs02099650_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price 4,505.00
          Your Price
          Online offer:
          4505.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          440073

          Gene Name:

          IQ motif and Sec7 domain 3

          Gene Aliases:

          -

          Location:

          Chr.12:66794-178460 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 15 - Intron 15
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          IQSEC3 NM_001170738.1 NP_001164209.1
          NM_015232.1 NP_056047.1
          XM_011520958.2 XP_011519260.1
          XM_011520959.2 XP_011519261.1
          XM_011520960.1 XP_011519262.1
          XM_011520961.1 XP_011519263.1
          XM_017019310.1 XP_016874799.1
          XM_017019311.1 XP_016874800.1
          AB029033.1 BAA83062.1
          AK091953.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv832302 Chr.12:57392 - 234835 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC574538 LOC101929384 SLC6A13
          esv3628226 Chr.12:157297 - 251505 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC101929384 SLC6A13
          nsv556853 Chr.12:136169 - 201168 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC574538
          nsv556854 Chr.12:152875 - 201168 on Build GRCh38 Loss IQSEC3 SLC6A12
          dgv87n21 Chr.12:151516 - 194154 on Build GRCh38 Loss IQSEC3 SLC6A12
          nsv1035539 Chr.12:119253 - 224079 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC574538 LOC101929384 SLC6A13
          nsv556855 Chr.12:154673 - 226756 on Build GRCh38 Loss IQSEC3 SLC6A12 LOC101929384 SLC6A13
          dgv1331n100 Chr.12:148770 - 207194 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC574538 LOC101929384
          nsv470252 Chr.12:84652 - 194154 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC574538
          nsv556856 Chr.12:170292 - 218255 on Build GRCh38 Loss IQSEC3 SLC6A12 LOC101929384
          esv3628221 Chr.12:114421 - 232803 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC574538 LOC101929384 SLC6A13
          nsv568 Chr.12:145682 - 179720 on Build GRCh38 Insertion IQSEC3 LOC574538
          dgv1330n100 Chr.12:45001 - 233978 on Build GRCh38 Gain IQSEC3 SLC6A12 LOC574538 LOC101929384 SLC6A13

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs76461415] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton organization
          regulation of ARF protein signal transduction
          positive regulation of GTPase activity
          ARF guanyl-nucleotide exchange factor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0