Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02215244_cn
          See other OR51E2 CNV Assays ›
          Gene Symbol
          OR51E2
          Assay Reference Genome
          Location

          Chr.11:4680812 on build GRCh38
          Cytoband
          11p15.4
          Assay ID Hs02215244_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          81285

          Gene Name:

          olfactory receptor family 51 subfamily E member 2

          Gene Aliases:

          OR51E3P, OR52A2, PSGR

          Location:

          Chr.11:4680171-4697846 on Build GRCh38

          Assay Gene Location:

          Within Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          OR51E2 NM_030774.3 2 2144 NP_110401.1
          AY033942.1 2 2141 AAK57550.1
          BC020768.1 4 1584 AAH20768.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3891914 Chr.11:4642313 - 4974456 on Build GRCh38 Gain OR51T1 OR51A2 OR51E2 OR51G2 OR51A4 OR52R1 OR51F1 OR51E1 OR51G1 OR51A7 OR51F2 OR51S1
          nsv1054847 Chr.11:4621645 - 5179426 on Build GRCh38 Gain OR52A5 OR51A2 OR52R1 OR51G1 MMP26 OR51A7 OR51F2 OR51S1 OR52E2 OR51D1 OR51T1 OR52J3 OR51E2 OR51G2 OR51A4 OR51F1 OR51E1 OR52A1 OR51L1 OR52A4P
          nsv832056 Chr.11:4651771 - 4803660 on Build GRCh38 Gain OR51E2 OR52R1 OR51F1 OR51E1
          nsv832055 Chr.11:4529664 - 4715110 on Build GRCh38 Loss OR51E2 OR52M1 OR51E1 OR52I1 TRIM68 OR52I2 C11orf40 OR51D1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          steroid hormone mediated signaling pathway
          positive regulation of blood pressure
          detection of chemical stimulus involved in sensory perception of smell
          cellular response to fatty acid
          positive regulation of renin secretion into blood stream
          steroid hormone receptor activity
          G-protein coupled receptor activity
          olfactory receptor activity
          signaling receptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline