Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02236423_cn
          See other PPOX CNV Assays ›
          Gene Symbol
          PPOX
          Assay Reference Genome
          Location

          Chr.1:161166644 on build GRCh38
          Cytoband
          Assay ID Hs02236423_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5498

          Gene Name:

          protoporphyrinogen oxidase

          Gene Aliases:

          PPO, V290M, VP, VPCO

          Location:

          Chr.1:161165728-161178013 on Build GRCh38

          Assay Gene Location:

          Within Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PPOX NM_000309.5 NP_000300.1
          NM_001122764.3 NP_001116236.1
          NM_001350128.2 NP_001337057.1
          NM_001350129.2 NP_001337058.1
          NM_001350130.2 NP_001337059.1
          NM_001350131.2 NP_001337060.1
          NM_001365398.1 NP_001352327.1
          NM_001365399.1 NP_001352328.1
          NM_001365400.1 NP_001352329.1
          NM_001365401.1 NP_001352330.1
          XM_005245291.5 XP_005245348.2
          XM_006711404.5 XP_006711467.3
          XM_011509663.3 XP_011507965.1
          XM_011509664.2 XP_011507966.1
          XM_011509665.3 XP_011507967.1
          XM_011509667.3 XP_011507969.1
          XM_011509668.3 XP_011507970.1
          XM_011509670.3 XP_011507972.1
          XM_011509671.2 XP_011507973.1
          XM_011509672.4 XP_011507974.2
          XM_011509673.3 XP_011507975.1
          XM_011509674.3 XP_011507976.1
          XM_024447864.2 XP_024303632.2
          XM_047423569.1 XP_047279525.1
          XM_047423581.1 XP_047279537.1
          AK094855.1 BAG52942.1
          AK298912.1 BAG61019.1
          AK302298.1 BAG63638.1
          AU076582.1
          BC002357.2 AAH02357.1
          BX384814.2
          D38537.1 BAA07538.1
          DA268550.1
          DA745403.1
          DC329287.1
          DC399187.1
          EL954312.1
          U26446.1 AAA67690.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6332269 Chr.1:160388767 - 162811116 on Build GRCh38 Duplication DDR2 VANGL2 SH2D1B APOA2 SPATA46 MIR4654 FCGR2C NDUFS2 PFDN2 DUSP12 B4GALT3 OLFML2B NIT1 FCGR2A ITLN2 FCGR3A SDHC CD244 MIR5187 SLAMF6 HSPA6 NOS1AP ITLN1 RPL31P11 TOMM40L KLHDC9 UAP1 F11R CD48 LOC112543491 LY9 LOC124904439 ATF6 SLAMF7 LOC100422526 CD84 FCER1G USP21 NECTIN4-AS1 FCRLB ATF6-DT PPOX DEDD HSPA7 USF1 FCGR2B C1orf226 FCGR3B LOC101928372 UFC1 CFAP126 CD84-AS1 UHMK1 MPZ ADAMTS4 FCRLA NR1I3 TSTD1 LOC105371470 HSD17B7 PCP4L1 NECTIN4 ARHGAP30 MIR556 SLAMF1
          nsv4518029 Chr.1:160848909 - 161416210 on Build GRCh38 Duplication PPOX DEDD APOA2 NDUFS2 USF1 PFDN2 B4GALT3 NIT1 LOC101928372 ITLN2 UFC1 SDHC CD244 CFAP126 MIR5187 MPZ ITLN1 TOMM40L ADAMTS4 NR1I3 KLHDC9 F11R LOC112543491 FCER1G USP21 NECTIN4-AS1 TSTD1 PCP4L1 NECTIN4 ARHGAP30

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          porphyrin-containing compound biosynthetic process
          protoporphyrinogen IX biosynthetic process
          heme biosynthetic process
          heme a biosynthetic process
          heme B biosynthetic process
          response to xenobiotic stimulus
          protoporphyrinogen IX metabolic process
          heme O biosynthetic process
          oxygen-dependent protoporphyrinogen oxidase activity
          oxidoreductase activity
          flavin adenine dinucleotide binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline