Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02294138_cn
          See other RUNX1T1 CNV Assays ›
          Gene Symbol
          RUNX1T1
          Assay Reference Genome
          Location

          Chr.8:91959895 on build GRCh38
          Cytoband
          Assay ID Hs02294138_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          862

          Gene Name:

          RUNX1 partner transcriptional co-repressor 1

          Gene Aliases:

          AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2

          Location:

          Chr.8:91954967-92103385 on Build GRCh38

          Assay Gene Location:

          Within Exon 18
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RUNX1T1 NM_001198625.2 15 2665 NP_001185554.1
          NM_001198626.2 13 2523 NP_001185555.1
          NM_001198627.2 14 2583 NP_001185556.1
          NM_001198628.2 12 2356 NP_001185557.1
          NM_001198629.2 12 2450 NP_001185558.1
          NM_001198630.2 13 2510 NP_001185559.1
          NM_001198631.2 12 2379 NP_001185560.1
          NM_001198632.2 12 2441 NP_001185561.1
          NM_001198633.2 11 2603 NP_001185562.1
          NM_001198634.2 11 2206 NP_001185563.1
          NM_001198679.3 12 2505 NP_001185608.1
          NM_001395209.1 12 2747 NP_001382138.1
          NM_004349.4 11 2163 NP_004340.1
          NM_175634.3 12 2776 NP_783552.1
          NM_175635.3 11 2277 NP_783553.1
          NM_175636.2 10 2391 NP_783554.1
          XM_011517351.3 13 2594 XP_011515653.1
          XM_017013936.2 11 2158 XP_016869425.1
          XM_017013939.2 7 2141 XP_016869428.1
          XM_017013940.2 8 3370 XP_016869429.1
          XM_017013941.2 7 3160 XP_016869430.1
          XM_024447318.2 11 2183 XP_024303086.1
          XM_047422373.1 12 7932 XP_047278329.1
          BC005850.1 11 2271 AAH05850.1
          BC067078.1 11 2262 AAH67078.2
          D14289.1 10 2348 BAA03247.1
          D14821.1 11 2246 BAA03558.1
          D43638.1 11 2492 BAA07755.1
          X79990.1 10 2074 CAA56311.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1022473 Chr.8:91854892 - 94250166 on Build GRCh38 Loss MIR378D2HG RBM12B RUNX1T1 RBM12B-AS1 GEM TMEM67 LOC105375638 MIR378D2 LOC101926956 CIBAR1-DT TRIQK LINC02906 MIR8084 CIBAR1 PDP1 CDH17 RBM12B-DT LOC102724710 FLJ46284

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          negative regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          DNA binding
          transcription corepressor activity
          protein binding
          zinc ion binding
          metal ion binding
          DNA-binding transcription factor binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline