Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02357645_cn
          See other FAM149A CNV Assays ›
          Gene Symbol
          FAM149A
          Assay Reference Genome
          Location

          Chr.4:186151987 on build GRCh38
          Cytoband
          Assay ID Hs02357645_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          25854

          Gene Name:

          family with sequence similarity 149 member A

          Gene Aliases:

          MST119, MSTP119

          Location:

          Chr.4:186104704-186175337 on Build GRCh38

          Assay Gene Location:

          Within Exon 6
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          FAM149A NM_001006655.3 4 612 NP_001006656.1
          NM_001350178.1 4 568 NP_001337107.1
          NM_001350179.1 4 541 NP_001337108.1
          NM_001367768.3 4 1247 NP_001354697.1
          NM_001395294.1 4 1274 NP_001382223.1
          NM_015398.4 4 561 NP_056213.1
          AF173894.1 3 243 AAQ13641.1
          AK057166.1 4 437 BAG51873.1
          AK295340.1 4 562 BAG58311.1
          AK297679.1 4 547 BAH12648.1
          AK309233.1 4 569
          AK311462.1
          BC110816.1 4 509 AAI10817.1
          BC136792.1 1 53 AAI36793.1
          BC142992.1 4 561 AAI42993.1
          BC144411.1 1 53 AAI44412.1
          BQ884007.1 4 589
          DA159632.1 4 477

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3894090 Chr.4:186011626 - 186212421 on Build GRCh38 Gain KLKB1 FAM149A TLR3 CYP4V2 FLJ38576
          nsv1016120 Chr.4:186125183 - 186169324 on Build GRCh38 Gain FAM149A
          nsv1028146 Chr.4:185582745 - 186417181 on Build GRCh38 Gain SORBS2 SORBS2-AS1 KLKB1 FAM149A F11 TLR3 CYP4V2 F11-AS1 FLJ38576
          nsv508340 Chr.4:186138019 - 186215804 on Build GRCh38 Deletion KLKB1 FAM149A CYP4V2 FLJ38576
          nsv6140977 Chr.4:186008703 - 186213068 on Build GRCh38 Duplication KLKB1 FAM149A TLR3 CYP4V2 FLJ38576
          nsv596401 Chr.4:186151229 - 186445495 on Build GRCh38 Loss KLKB1 FAM149A F11 CYP4V2 F11-AS1 FLJ38576
          nsv470097 Chr.4:186010756 - 186210350 on Build GRCh38 Gain FAM149A TLR3 CYP4V2 FLJ38576
          nsv4117959 Chr.4:185482576 - 186997201 on Build GRCh38 Deletion SORBS2 SORBS2-AS1 PDLIM3 KLKB1 FAT1 MTNR1A FAM149A F11 TLR3 CYP4V2 F11-AS1 FLJ38576
          nsv596400 Chr.4:186145486 - 186171321 on Build GRCh38 Loss FAM149A
          nsv4132364 Chr.4:186040783 - 186195278 on Build GRCh38 Deletion FAM149A TLR3 CYP4V2 FLJ38576
          nsv830172 Chr.4:186091611 - 186247270 on Build GRCh38 Loss KLKB1 FAM149A CYP4V2 FLJ38576
          dgv683n27 Chr.4:186008072 - 186210350 on Build GRCh38 Gain FAM149A TLR3 CYP4V2 FLJ38576
          nsv4521971 Chr.4:186115844 - 186466846 on Build GRCh38 Duplication KLKB1 FAM149A F11 CYP4V2 F11-AS1 FLJ38576
          nsv461864 Chr.4:186085652 - 186335850 on Build GRCh38 Gain KLKB1 FAM149A F11 TLR3 CYP4V2 F11-AS1 FLJ38576
          dgv1052e214 Chr.4:186072002 - 186212619 on Build GRCh38 Gain KLKB1 FAM149A TLR3 CYP4V2 FLJ38576

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline