Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02386801_cn
          See other ZNF229 CNV Assays ›
          Gene Symbol
          ZNF229
          Assay Reference Genome
          Location

          Chr.19:44430120 on build GRCh38
          Cytoband
          19q13.31
          Assay ID Hs02386801_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          7772

          Gene Name:

          zinc finger protein 229

          Gene Aliases:

          -

          Location:

          Chr.19:44426251-44448578 on Build GRCh38

          Assay Gene Location:

          Within Exon 6
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ZNF229 NM_001278510.2 6 966 NP_001265439.2
          NM_014518.3 6 1095 NP_055333.3
          NR_103551.2 6 1550
          XM_006723372.3 6 1169 XP_006723435.1
          XM_011527292.1 6 1077 XP_011525594.1
          XM_017027280.1 6 1624 XP_016882769.1
          AB209357.1 6 1519 BAD92594.1
          AF192979.1 6 1096 AAF07964.1
          BC150612.1 4 831
          CR936724.1 6 967

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3644482 Chr.19:44388164 - 44434579 on Build GRCh38 Loss ZNF285 ZNF229
          nsv1061350 Chr.19:44364917 - 44531444 on Build GRCh38 Loss ZNF180 ZNF285 ZNF229 CEACAM20
          esv2663330 Chr.19:44388193 - 44434570 on Build GRCh38 Deletion ZNF285 ZNF229
          nsv817849 Chr.19:44398850 - 44430844 on Build GRCh38 Loss ZNF285 ZNF229
          esv2763206 Chr.19:44381218 - 44437545 on Build GRCh38 Loss ZNF285 ZNF229
          nsv579857 Chr.19:44424713 - 44430844 on Build GRCh38 Loss ZNF229
          nsv515911 Chr.19:44347982 - 44430844 on Build GRCh38 Loss ZNF285 ZNF112 ZNF229
          esv2421963 Chr.19:44392234 - 44434616 on Build GRCh38 Deletion ZNF285 ZNF229
          nsv9734 Chr.19:44388599 - 44435840 on Build GRCh38 Gain+Loss ZNF285 ZNF229
          esv2758764 Chr.19:44389939 - 44443260 on Build GRCh38 Loss ZNF285 ZNF229
          nsv1062330 Chr.19:44359787 - 44434102 on Build GRCh38 Loss ZNF285 ZNF229
          dgv3605n100 Chr.19:44390001 - 44434604 on Build GRCh38 Loss ZNF285 ZNF229
          esv26773 Chr.19:44424072 - 44434546 on Build GRCh38 Loss ZNF229

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline