Search
Search

Entrez Gene ID: | 80173 |
Gene Name: | intraflagellar transport 74 |
Gene Aliases: |
CCDC2, CMG-1, CMG1 |
Location: |
Chr.9:26947039-27066129 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT74 | NM_001099222.1 | 9 | 813 | NP_001092692.1 |
| NM_001099223.1 | 9 | 769 | NP_001092693.1 | |
| NM_001099224.1 | 9 | 805 | NP_001092694.1 | |
| NM_025103.2 | 9 | 805 | NP_079379.2 | |
| XM_011518035.2 | 9 | 805 | XP_011516337.1 | |
| XM_011518036.2 | 2 | 515 | XP_011516338.1 | |
| XM_017015163.1 | 2 | 320 | XP_016870652.1 | |
| XM_017015164.1 | 2 | 1761 | XP_016870653.1 | |
| AK023707.1 | 9 | 766 | BAB14650.1 | |
| AK225570.1 | 9 | 766 | ||
| AY040325.1 | 9 | 734 | AAK77221.1 | |
| BC023634.2 | 9 | 750 | ||
| BC107742.1 | 9 | 703 | AAI07743.1 | |
| BC113524.1 | 8 | 578 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517269 | Chr.9:26945978 - 27137244 on Build GRCh38 | Gain |
|
| nsv528614 | Chr.9:26893664 - 27074235 on Build GRCh38 | Gain |
|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
| esv3891647 | Chr.9:26743024 - 27123262 on Build GRCh38 | Gain |
|
Set Membership: |
|