Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs02510167_cn
See other MDM2 CNV Assays ›
Gene Symbol
MDM2
Assay Reference Genome
Location
Chr.12:68820374 on build GRCh38
Cytoband
12q15
Assay ID
Hs02510167_cn
Size
S: 360 rxns
M: 750 rxns
L: 2,900 rxns
Availability
Made To Order
Catalog #
4400291
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Target Gene Details
Entrez Gene ID:
4193
Gene Name:
MDM2 proto-oncogene
Gene Aliases:
ACTFS, HDMX, hdm2
Location:
Chr.12:68808149-68845544 on Build GRCh38
Assay Gene Location:
Overlaps Exon 7 - Intron 7
Gene Symbol
Transcript Accession
Exon Location
Assay Transcript Location
Protein ID
MDM2
NM_001145337.2
NP_001138809.1
NM_001145339.2
NP_001138811.1
NM_001145340.2
NP_001138812.1
NM_001278462.1
NP_001265391.1
NM_002392.5
NP_002383.2
XM_005268872.4
XP_005268929.1
XM_006719399.3
XP_006719462.1
XM_006719400.3
XP_006719463.1
AF092843.1
AAL40178.1
AF092844.1
AAL40179.1
AF092845.1
AAL40180.1
AF201370.1
AAF42995.1
AF201371.1
AF385322.1
AAL13242.1
AF385323.1
AAL13243.1
AF385324.1
AAL13244.1
AF385325.1
AAL13245.1
AF385326.1
AAL13246.1
AF385327.1
AAL13247.1
AJ276888.1
AJ278975.1
CAC07809.1
AJ278976.1
CAC07810.1
AJ278977.1
CAC07811.1
AJ278978.1
CAC07812.1
AJ430612.1
CAD23251.1
AJ430613.1
AJ430614.1
CAD23252.1
AJ491698.1
CAD36959.1
AJ491699.1
CAD36960.1
AJ491700.1
CAD36961.1
AJ550516.1
CAD79455.1
AJ550517.1
CAD79456.1
AJ550518.1
CAD79457.1
AJ550519.1
CAD79458.1
AJ550520.1
CAD79459.1
AK290341.1
AM902641.1
AM902642.1
AM902643.1
AM902644.1
AM902645.1
AM902646.1
AM902647.1
AM902648.1
AM902649.1
AM902650.1
AM902651.1
AM902652.1
AM902653.1
AM902654.1
AM902655.1
AM902656.1
AM902657.1
AM902658.1
AM902659.1
AM902660.1
AM902661.1
AM902662.1
AM902663.1
AM902664.1
AM902665.1
AM902666.1
AM902667.1
AM902668.1
AM902669.1
AM902670.1
AM902671.1
AM902672.1
AM902673.1
AM902674.1
AM902675.1
AM902676.1
AM902677.1
AM902678.1
AM902679.1
AM902680.1
AM902681.1
BC152384.1
BC152390.1
BT007258.1
AAP35922.1
EU076746.1
EU076747.1
EU076748.1
EU076749.1
GQ848196.1
HM213901.1
HM213902.1
HM213903.1
HM213904.1
JX015379.1
JX015380.1
JX015381.1
JX015382.1
JX015383.1
JX015384.1
JX015385.1
JX015387.1
JX015388.1
JX015389.1
JX015390.1
JX015391.1
JX015392.1
M92424.1
AAA60568.1
U33199.1
AAA75514.1
U33200.1
AAA75515.1
U33201.1
AAA75516.1
U33202.1
AAA75517.1
U33203.1
AAA75518.1
Z12020.1
CAA78055.1
Back To Top
More Information
Set Membership:
Intragenic
Non-exonic
non-DGV Variation
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
blood vessel remodeling
regulation of heart rate
atrioventricular valve morphogenesis
endocardial cushion morphogenesis
ventricular septum development
atrial septum development
protein complex assembly
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
traversing start control point of mitotic cell cycle
positive regulation of cell proliferation
response to toxic substance
response to carbohydrate
response to iron ion
positive regulation of gene expression
negative regulation of protein processing
viral process
protein ubiquitination
protein sumoylation
peptidyl-lysine modification
protein destabilization
response to magnesium ion
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein localization to nucleus
regulation of protein catabolic process
response to cocaine
response to drug
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to morphine
negative regulation of DNA damage response, signal transduction by p53 class mediator
establishment of protein localization
response to ether
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
response to antibiotic
positive regulation of protein export from nucleus
response to steroid hormone
cardiac septum morphogenesis
cellular response to hydrogen peroxide
negative regulation of cell cycle arrest
cellular response to antibiotic
cellular response to vitamin B1
cellular response to alkaloid
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to estrogen stimulus
cellular response to hypoxia
cellular response to UV-C
regulation of signal transduction by p53 class mediator
negative regulation of signal transduction by p53 class mediator
p53 binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
SUMO transferase activity
enzyme binding
ubiquitin protein ligase binding
identical protein binding
peroxisome proliferator activated receptor binding
ubiquitin protein ligase activity
scaffold protein binding
Back To Top
Related Products
TaqMan
®
Genotyping Master Mix
TaqPath
®
ProAmp
®
Master Mix
TaqMan
®
Universal Master Mix II, no UNG
TaqMan
®
Gene Expression Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Universal PCR Master Mix, No AmpErase UNG
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline