Search
Search

Entrez Gene ID: | 7738 |
Gene Name: | zinc finger protein 184 |
Gene Aliases: |
kr-ZNF3 |
Location: |
Chr.6:27404006-27473105 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF184 | NM_001318891.2 | 6 | 2365 | NP_001305820.1 |
| NM_001318892.2 | 6 | 2433 | NP_001305821.1 | |
| NM_001318893.2 | 6 | 2394 | NP_001305822.1 | |
| NM_001347832.2 | 6 | 2150 | NP_001334761.1 | |
| NM_007149.3 | 6 | 2121 | NP_009080.2 | |
| NR_144687.2 | ||||
| NR_144688.2 | ||||
| AK123011.1 | 5 | 2660 | ||
| AK131264.1 | 7 | 2134 | BAD18442.1 | |
| AK312804.1 | 6 | 2375 | BAG35662.1 | |
| BC022992.1 | 6 | 2116 | AAH22992.1 | |
| DA315543.1 | ||||
| U66561.1 | 3 | 1774 | AAC51180.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6398741 | Chr.6:27314958 - 28461291 on Build GRCh38 | Duplication |
|
Set Membership: |
|