Search
Search

Entrez Gene ID: | 1822 |
Gene Name: | atrophin 1 |
Gene Aliases: |
B37, D12S755E, DRPLA, HRS, NOD |
Location: |
Chr.12:6924463-6942321 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATN1 | NM_001007026.1 | NP_001007027.1 | ||
| NM_001940.3 | NP_001931.2 | |||
| BC051795.2 | AAH51795.1 | |||
| BQ880124.1 | ||||
| D31840.1 | BAA06626.1 | |||
| D38529.1 | BAA07534.1 | |||
| U23851.1 | AAB50276.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain |
|
| nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion |
|
| nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain |
|
Set Membership: |
|