Search
Search

Entrez Gene ID: | 861 |
Gene Name: | RUNX family transcription factor 1 |
Gene Aliases: |
AML1, AML1-EVI-1, AMLCR1, CBF2alpha, CBFA2, EVI-1, PEBP2aB, PEBP2alpha |
Location: |
Chr.21:34787801-35049302 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNX1 | NM_001001890.3 | 6 | 6126 | NP_001001890.1 |
| NM_001754.5 | 9 | 4814 | NP_001745.2 | |
| XM_005261068.4 | 7 | 4607 | XP_005261125.1 | |
| XM_005261069.5 | 8 | 4622 | XP_005261126.1 | |
| XM_011529766.3 | 9 | 4901 | XP_011528068.1 | |
| XM_011529767.3 | 8 | 4775 | XP_011528069.1 | |
| XM_011529768.3 | 7 | 4583 | XP_011528070.1 | |
| XM_047441007.1 | 8 | 5023 | XP_047296963.1 | |
| XM_047441009.1 | 7 | 4984 | XP_047296965.1 | |
| XM_047441010.1 | 8 | 4862 | XP_047296966.1 | |
| XM_047441011.1 | 7 | 4831 | XP_047296967.1 | |
| XM_047441012.1 | 8 | 4709 | XP_047296968.1 | |
| XM_047441013.1 | 6 | 4415 | XP_047296969.1 | |
| XM_047441014.1 | 6 | 4792 | XP_047296970.1 | |
| XM_047441015.1 | 7 | 4670 | XP_047296971.1 | |
| CD638998.1 | 1 | 201 | ||
| D43968.1 | 6 | 6117 | BAA07903.1 | |
| D43969.1 | 8 | 5064 | BAA07904.1 | |
| X90976.1 | 1 | 3222 | CAA62464.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5550749 | Chr.21:34567808 - 34891808 on Build GRCh38 | Duplication |
|
| nsv5539790 | Chr.21:5227438 - 39583816 on Build GRCh38 | Insertion |
|
Set Membership: |
|