Search
Search

Entrez Gene ID: | 57644 |
Gene Name: | myosin heavy chain 7B |
Gene Aliases: |
MHC14, MYH14, lncMYH7b |
Location: |
Chr.20:34955868-35002437 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 33 - Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYH7B | NM_020884.7 | NP_065935.4 | ||
| XM_006723840.4 | XP_006723903.1 | |||
| XM_047440336.1 | XP_047296292.1 | |||
| XM_047440337.1 | XP_047296293.1 | |||
| XM_047440338.1 | XP_047296294.1 | |||
| XM_047440339.1 | XP_047296295.1 | |||
| XM_047440340.1 | XP_047296296.1 | |||
| XM_047440341.1 | XP_047296297.1 | |||
| AB040945.2 | BAA96036.3 | |||
| BC007808.1 | AAH07808.1 | |||
| BC151242.1 | AAI51243.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063673 | Chr.20:34977862 - 35009073 on Build GRCh38 | Loss |
|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
| nsv833962 | Chr.20:34927688 - 35113250 on Build GRCh38 | Loss |
|
| nsv3362 | Chr.20:34978502 - 35010725 on Build GRCh38 | Insertion |
|
Set Membership: |
|