Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02708460_cn
          See other OR5D14 CNV Assays ›
          Gene Symbol
          OR5D14
          Assay Reference Genome
          Location

          Chr.11:55796015 on build GRCh38
          Cytoband
          11q11
          Assay ID Hs02708460_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          219436

          Gene Name:

          olfactory receptor family 5 subfamily D member 14

          Gene Aliases:

          OR11-141, OR11-150

          Location:

          Chr.11:55795556-55796500 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          OR5D14 NM_001004735.1 1 460 NP_001004735.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2751030 Chr.11:55068147 - 56111950 on Build GRCh38 Gain OR5L1 OR4C1P OR5AS1 TRIM51HP OR8I2 OR4S2 OR7E5P OR4C15 OR5I1 OR4C6 OR5F1 TRIM51 OR4C16 OR8H2 OR5D14 OR4A15 OR4P4 OR5L2 OR5D18 OR5F2P TRIM48 OR4A16 OR5W2 OR10AG1 OR5D13 OR5D16 OR4C11
          nsv437717 Chr.11:55592180 - 55839217 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR5D16 OR4C11
          nsv554801 Chr.11:55546675 - 55856884 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR5D16 OR4C11 OR4C16 OR4C15
          nsv469958 Chr.11:55603544 - 55828837 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR4C11
          dgv1905n54 Chr.11:55267935 - 56096685 on Build GRCh38 Gain OR5L1 OR4C1P OR5AS1 TRIM51HP OR8I2 OR4S2 OR7E5P OR4C15 OR5I1 OR4C6 OR5F1 TRIM51 OR4C16 OR5D14 OR4A15 OR4P4 OR5L2 OR5D18 OR5F2P TRIM48 OR4A16 OR5W2 OR10AG1 OR5D13 OR5D16 OR4C11
          dgv1205n100 Chr.11:55693312 - 56247291 on Build GRCh38 Gain OR5D14 OR5L2 OR5L1 OR5AS1 OR5D18 OR8I2 OR5F2P OR5W2 OR8K5 OR7E5P OR8J3 OR5I1 OR5T2 OR10AG1 OR8H3 OR5D13 OR5F1 OR5D16 TRIM51 OR8H2 OR5J2
          esv2760216 Chr.11:55591984 - 55842207 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR5D16 OR4C11
          dgv51e55 Chr.11:55677818 - 55853223 on Build GRCh38 Loss OR5D14 OR5L2 OR5L1 OR5D13 OR5D18 OR5D16
          dgv212e199 Chr.11:55654858 - 55836170 on Build GRCh38 Deletion OR5D14 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6
          esv3579782 Chr.11:55686087 - 56244978 on Build GRCh38 Gain OR5D14 OR5L2 OR5L1 OR5AS1 OR5D18 OR8I2 OR5F2P OR5W2 OR8K5 OR7E5P OR8J3 OR5I1 OR5T2 OR10AG1 OR8H3 OR5D13 OR5F1 OR5D16 TRIM51 OR8H2 OR5J2
          nsv832158 Chr.11:55782684 - 55933833 on Build GRCh38 Gain OR5D14 OR5L2 OR5L1 OR5D18 OR5D16 TRIM51 OR5W2
          dgv50e55 Chr.11:55609530 - 55853223 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR5D16
          dgv13n68 Chr.11:55615204 - 55826897 on Build GRCh38 Loss OR5D14 OR4P4 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2
          nsv1044174 Chr.11:55734320 - 56094696 on Build GRCh38 Loss OR5D14 OR5L2 OR5L1 OR5AS1 OR5D18 OR8I2 OR5F2P OR5W2 OR7E5P OR5I1 OR10AG1 OR5D13 OR5F1 OR5D16 TRIM51
          dgv1941n54 Chr.11:55679951 - 55836936 on Build GRCh38 Loss OR5D14 OR5L2 OR5L1 OR5D13 OR5D18
          nsv428256 Chr.11:55549600 - 55854408 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR5D16 OR4C11 OR4C16 OR4C15
          esv2756713 Chr.11:55696306 - 55816633 on Build GRCh38 Loss OR5D14 OR5L1 OR5D13
          dgv1204n100 Chr.11:55654851 - 55865297 on Build GRCh38 Loss OR5D14 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR5D16
          nsv517440 Chr.11:55572903 - 55836936 on Build GRCh38 Gain+Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR4C11 OR4C16
          nsv821668 Chr.11:55386454 - 56049352 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR4C1P OR5AS1 OR5D18 OR5F2P OR4S2 OR5W2 OR7E5P OR4C15 OR5I1 OR10AG1 OR5D13 OR4C6 OR5F1 OR5D16 TRIM51 OR4C11 OR4C16
          esv22842 Chr.11:55597201 - 55836114 on Build GRCh38 Gain+Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2 OR4C11
          nsv469885 Chr.11:55678817 - 55851147 on Build GRCh38 Loss OR5D14 OR5L2 OR5L1 OR5D13 OR5D18 OR5D16
          esv2759828 Chr.11:55454944 - 55870636 on Build GRCh38 Gain+Loss OR5D14 OR4P4 OR5L2 OR5L1 OR4C1P OR5D18 OR4S2 OR4C15 OR5D13 OR4C6 OR5D16 OR4C11 OR4C16
          dgv12n64 Chr.11:55655110 - 55828837 on Build GRCh38 Loss OR5D14 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6
          dgv1193n100 Chr.11:55609010 - 55836203 on Build GRCh38 Loss OR5D14 OR4P4 OR5L2 OR5L1 OR5D13 OR5D18 OR4C6 OR4S2
          esv3626453 Chr.11:55691646 - 55841751 on Build GRCh38 Loss OR5D14 OR5L2 OR5L1 OR5D13 OR5D18 OR5D16

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline