Search
Search

Entrez Gene ID: | 5554 |
Gene Name: | proline rich protein HaeIII subfamily 1 |
Gene Aliases: |
Db-s, PA, PIF-S, PRH2, PRP-1/PRP-2, Pr1/Pr2 |
Location: |
Chr.12:79671-366551 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH1 | NM_001291314.2 | NP_001278243.1 | ||
| NM_001291315.2 | NP_001278244.1 | |||
| AB593029.1 | BAJ83983.1 | |||
| BI772091.1 | ||||
| BM476484.1 | ||||
| BU543118.1 |
Entrez Gene ID: | 100533464 |
Gene Name: | PRH1-PRR4 readthrough |
Gene Aliases: |
PRH1 |
Location: |
Chr.12:44555-366565 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH1-PRR4 | NR_037918.2 | |||
| AB593030.1 | BAJ83984.1 |
Entrez Gene ID: | 5555 |
Gene Name: | proline rich protein HaeIII subfamily 2 |
Gene Aliases: |
PRP-1/PRP-2, Pr, pr1/Pr2 |
Location: |
Chr.12:108262-133551 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH2 | NM_001110213.1 | NP_001103683.1 | ||
| XM_047429101.1 | 3 | 1467 | XP_047285057.1 | |
| BC095488.1 | AAH95488.1 | |||
| BC136499.1 | 3 | 1458 | AAI36500.1 | |
| BX538206.1 | ||||
| BX538207.1 | ||||
| BX641094.1 | 3 | 1471 | CAE46044.1 | |
| BX647085.1 | ||||
| BX647155.1 | 4 | 1365 | ||
| BX647156.1 | ||||
| BX648436.1 | ||||
| BX648656.1 | 3 | 1397 | ||
| BX648871.1 | 2 | 1442 | ||
| BX648901.1 | ||||
| CR749583.1 | CAH18378.1 | |||
| K03202.1 | AAA60183.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5494258 | Chr.12:10925220 - 10950133 on Build GRCh38 | Deletion |
|
| nsv6465694 | Chr.12:10923438 - 10933265 on Build GRCh38 | Deletion |
|
| nsv832330 | Chr.12:10925079 - 11054358 on Build GRCh38 | Gain |
|
| esv2759879 | Chr.12:10817021 - 11561987 on Build GRCh38 | Loss |
|
| nsv1038277 | Chr.12:10908379 - 11351157 on Build GRCh38 | Loss |
|
| nsv428272 | Chr.12:10917246 - 11561987 on Build GRCh38 | Gain+Loss |
|
| esv3628550 | Chr.12:10925203 - 10950111 on Build GRCh38 | Loss |
|
Set Membership: |
|