Search
Search

Entrez Gene ID: | 8659 |
Gene Name: | aldehyde dehydrogenase 4 family member A1 |
Gene Aliases: |
ALDH4, P5CD, P5CDh |
Location: |
Chr.1:18871430-18902555 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH4A1 | NM_001161504.2 | NP_001154976.1 | ||
| NM_001319218.2 | NP_001306147.1 | |||
| NM_003748.4 | NP_003739.2 | |||
| NM_170726.3 | NP_733844.1 | |||
| AK222486.1 | BAD96206.1 | |||
| AK289972.1 | BAF82661.1 | |||
| AK294552.1 | BAG57755.1 | |||
| BC007581.1 | AAH07581.1 | |||
| BC023600.2 | AAH23600.1 | |||
| BM856369.1 | ||||
| FJ462711.1 | ACN89883.1 | |||
| GQ891378.1 | ADO22240.1 | |||
| U24266.1 | AAC50500.1 | |||
| U24267.1 | AAC50501.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4032896 | Chr.1:18854016 - 18914915 on Build GRCh38 | Duplication |
|
Set Membership: |
|