Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02777838_cn
          See other TXNIP CNV Assays ›
          Gene Symbol
          TXNIP
          Assay Reference Genome
          Location

          Chr.1:145992646 on build GRCh38
          Cytoband
          Assay ID Hs02777838_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          10628

          Gene Name:

          thioredoxin interacting protein

          Gene Aliases:

          ARRDC6, EST01027, HHCPA78, THIF, VDUP1

          Location:

          Chr.1:145992435-145996579 on Build GRCh38

          Assay Gene Location:

          Within Exon 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TXNIP NM_001313972.2 7 2399 NP_001300901.1
          NM_006472.6 8 2694 NP_006463.3
          AA805348.1 1 234
          BQ030003.1 1 238
          BX404012.2 1 875
          BX537824.1 5 2128
          S73591.1 8 2602 AAB31977.2

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss GPR89A GPR89B PRKAB2 PDE4DIPP1 PPIAL4H BCL9 ACP6 MIR6736 GNRHR2 GJA8 NBPF14 SEC22B3P RNVU1-3 LINC02806 CD160 FMO5 GJA5 PDZK1 PDE4DIPP6 RNVU1-7 NOTCH2NLA NUDT4P2 ANKRD34A LOC728989 RNVU1-2 RNF115 LOC128071544 PDE4DIP RNVU1-1 NUDT17 NBPF11 LINC02805 NBPF13P HYDIN2 PPIAL4G MIR5087 TXNIP NOTCH2NLB NBPF12 RNVU1-8 PDIA3P1 LINC01731 LINC00624 CHD1L PDZK1P1 LIX1L-AS1 RNVU1-6 NUDT4B PEX11B HJV LOC101927468 RNVU1-27 ITGA10 LIX1L POLR3GL POLR3C NBPF10 ANKRD35 RBM8A PIAS3 MIR6077 LINC01719 LINC01138
          nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion GPR89B PRKAB2 PDE4DIPP1 PPIAL4H BCL9 ACP6 GJA8 NBPF14 SEC22B3P RNVU1-3 LINC02806 FMO5 GJA5 PDE4DIPP6 RNVU1-7 NOTCH2NLA NUDT4P2 ANKRD34A LOC728989 RNVU1-2 LOC128071544 PDE4DIP RNVU1-1 NBPF11 LINC02805 NBPF13P HYDIN2 PPIAL4G MIR5087 TXNIP NOTCH2NLB NBPF12 RNVU1-8 PDIA3P1 LINC01731 LINC00624 CHD1L PDZK1P1 RNVU1-6 NUDT4B HJV LOC101927468 RNVU1-27 LIX1L POLR3GL NBPF10 MIR6077 LINC01719 LINC01138
          nsv950948 Chr.1:145977587 - 146000259 on Build GRCh38 Duplication TXNIP POLR3GL
          nsv6138813 Chr.1:144966587 - 146310000 on Build GRCh38 Duplication LOC100996740 GPR89A NUDT17 TRN-GTT2-7 NBPF25P PPIAL4D TXNIP RNVU1-14 MIR6736 GNRHR2 FAM72D LOC101929805 SRGAP2B LIX1L-AS1 CD160 RNVU1-6 PEX11B HJV PDZK1 ITGA10 LIX1L POLR3GL NOTCH2NLA RNVU1-31 POLR3C NUDT4P2 NBPF10 ANKRD35 RBM8A LINC01145 ANKRD34A PIAS3 LINC01719 RNF115 NBPF20
          nsv5881371 Chr.1:145648753 - 147971330 on Build GRCh38 Deletion GPR89A GPR89B PRKAB2 NUDT17 NBPF13P HYDIN2 PPIAL4H BCL9 TXNIP ACP6 MIR6736 GNRHR2 GJA8 NBPF12 RNVU1-8 PDIA3P1 LINC00624 CHD1L LIX1L-AS1 CD160 FMO5 RNVU1-6 GJA5 PEX11B HJV PDZK1 ITGA10 LIX1L POLR3GL NOTCH2NLA POLR3C NUDT4P2 NBPF10 ANKRD35 RBM8A ANKRD34A LOC728989 PIAS3 LINC01719 RNF115 LOC128071544
          nsv831359 Chr.1:145983609 - 146147220 on Build GRCh38 Loss TXNIP RNVU1-6 NBPF10 HJV LINC01719
          nsv6333880 Chr.1:145488103 - 148061210 on Build GRCh38 Deletion GPR89A GPR89B PRKAB2 PDE4DIPP1 NUDT17 NBPF13P HYDIN2 NBPF25P PPIAL4H BCL9 TXNIP ACP6 MIR6736 GNRHR2 GJA8 NBPF12 RNVU1-8 PDIA3P1 LINC00624 CHD1L PDZK1P1 LIX1L-AS1 CD160 FMO5 RNVU1-6 GJA5 PEX11B HJV PDZK1 ITGA10 LIX1L POLR3GL RNVU1-7 NOTCH2NLA POLR3C NUDT4P2 NBPF10 ANKRD35 RBM8A ANKRD34A LOC728989 PIAS3 LINC01719 RNF115 LOC128071544

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein import into nucleus
          inflammatory response
          response to oxidative stress
          response to xenobiotic stimulus
          response to mechanical stimulus
          response to glucose
          protein transport
          keratinocyte differentiation
          response to estradiol
          response to progesterone
          regulation of cell proliferation
          response to hydrogen peroxide
          positive regulation of apoptotic process
          platelet-derived growth factor receptor signaling pathway
          response to calcium ion
          negative regulation of cell division
          cellular response to tumor cell
          cellular response to oxidised low-density lipoprotein particle stimulus
          enzyme inhibitor activity
          protein binding
          ubiquitin protein ligase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline