Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02951422_cn
          See other PIGB CNV Assays ›
          Gene Symbol
          PIGB
          Assay Reference Genome
          Location

          Chr.15:55319308 on build GRCh38
          Cytoband
          Assay ID Hs02951422_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9488

          Gene Name:

          phosphatidylinositol glycan anchor biosynthesis class B

          Gene Aliases:

          DEE80, EIEE80, GPI-MT-III, PIG-B

          Location:

          Chr.15:55319222-55355648 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PIGB NM_004855.5 1 87 NP_004846.4
          XM_005254795.6 1 87 XP_005254852.1
          XM_011522235.4 1 87 XP_011520537.1
          XM_011522236.4 1 87 XP_011520538.1
          XM_011522237.3 1 87 XP_011520539.1
          XM_047433363.1 1 87 XP_047289319.1
          XM_047433364.1 1 87 XP_047289320.1
          XM_047433365.1 1 87 XP_047289321.1
          AK223330.1 1 79 BAD97050.1
          AK293666.1 1 349 BAG57112.1
          AK314706.1 1 349 BAG37253.1
          BC017711.1 1 311 AAH17711.1
          D42138.1 1 102 BAA07709.1
          DA598474.1 1 374

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2761899 Chr.15:53592144 - 55643724 on Build GRCh38 Gain+Loss LOC105370829 MIR628 DNAAF4 DNAAF4-CCPG1 PIGBOS1 CCPG1 UNC13C RAB27A PIERCE2 PIGB RSL24D1 WDR72 PYGO1 PRTG LOC124903496
          nsv4242082 Chr.15:55003981 - 55380858 on Build GRCh38 Duplication LOC105370829 MIR628 RAB27A DNAAF4-CCPG1 PIGBOS1 CCPG1 PIGB RSL24D1
          nsv6515550 Chr.15:55306219 - 55474102 on Build GRCh38 Duplication MIR628 RAB27A DNAAF4 PIERCE2 DNAAF4-CCPG1 PIGBOS1 CCPG1 PIGB
          nsv1043789 Chr.15:55108674 - 55800256 on Build GRCh38 Gain MIR628 RAB27A DNAAF4 PIERCE2 DNAAF4-CCPG1 PIGBOS1 CCPG1 PIGB RSL24D1 PYGO1 PRTG LOC124903496
          nsv569523 Chr.15:55263404 - 55544506 on Build GRCh38 Gain MIR628 RAB27A DNAAF4 PIERCE2 DNAAF4-CCPG1 PIGBOS1 CCPG1 PIGB PYGO1 LOC124903496
          esv3569598 Chr.15:55270429 - 55410270 on Build GRCh38 Gain MIR628 RAB27A PIERCE2 DNAAF4-CCPG1 PIGBOS1 CCPG1 PIGB
          nsv1036924 Chr.15:55235676 - 55324282 on Build GRCh38 Loss RAB27A PIGBOS1 PIGB
          nsv569524 Chr.15:55266426 - 55320313 on Build GRCh38 Loss RAB27A PIGBOS1 PIGB
          nsv4247248 Chr.15:55307775 - 55493742 on Build GRCh38 Duplication MIR628 RAB27A DNAAF4 PIERCE2 DNAAF4-CCPG1 PIGBOS1 CCPG1 PIGB

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          alpha-1,2-mannosyltransferase activity
          glycolipid mannosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline