Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03186416_cn
          See other SLC2A14 CNV Assays ›
          Gene Symbol
          SLC2A14
          Assay Reference Genome
          Location

          Chr.12:7821266 on build GRCh38
          Cytoband
          12p13.31
          Assay ID Hs03186416_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          144195

          Gene Name:

          solute carrier family 2 member 14

          Gene Aliases:

          GLUT14, SLC2A3P3

          Location:

          Chr.12:7812512-7891196 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 16 - Exon 16
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SLC2A14 NM_001286233.1 NP_001273162.1
          NM_001286234.1 NP_001273163.1
          NM_001286235.1 NP_001273164.1
          NM_001286236.1 NP_001273165.1
          NM_001286237.1 NP_001273166.1
          NM_153449.3 NP_703150.1
          XM_005253315.3 XP_005253372.1
          XM_005253317.4 XP_005253374.1
          XM_011520562.1 XP_011518864.1
          XM_011520563.2 XP_011518865.1
          XM_011520564.2 XP_011518866.1
          XM_011520565.2 XP_011518867.1
          XM_017018841.1 XP_016874330.1
          XM_017018842.1 XP_016874331.1
          XM_017018843.1 XP_016874332.1
          XM_017018844.1 XP_016874333.1
          XM_017018845.1 XP_016874334.1
          XM_017018846.1 XP_016874335.1
          XM_017018847.1 XP_016874336.1
          AF481878.1 AAL89709.1
          AF481879.1 AAL89710.1
          AK122783.1
          AK126026.1
          AK299037.1
          AK302881.1
          AK316238.1
          AL110298.1 CAB53739.2
          AL832448.1
          AY357941.1 AAQ63763.1
          BC060766.1 AAH60766.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv58e55 Chr.12:7801455 - 7987805 on Build GRCh38 Gain SLC2A14 NANOGP1 SLC2A3
          esv2656717 Chr.12:7798646 - 7905603 on Build GRCh38 Deletion SLC2A14 NANOGP1
          esv2751114 Chr.12:7810616 - 7964473 on Build GRCh38 Loss SLC2A14 NANOGP1 SLC2A3
          esv2759877 Chr.12:7704223 - 8471391 on Build GRCh38 Gain+Loss CLEC6A C3AR1 SLC2A14 FAM66C FOXJ2 NECAP1 FAM86FP LINC00937 FAM90A1 LOC101927905 DPPA3 SLC2A3 NANOGP1 POU5F1P3 CLEC4A CLEC4C NANOGNB ZNF705A NANOG
          dgv2329n54 Chr.12:7808980 - 7993566 on Build GRCh38 Gain SLC2A14 NANOGP1 SLC2A3
          nsv1070090 Chr.12:7798719 - 7905639 on Build GRCh38 Deletion SLC2A14 NANOGP1
          esv2422393 Chr.12:7682970 - 7834994 on Build GRCh38 Duplication SLC2A14 GDF3 CLEC4C NANOGNB DPPA3 NANOG
          esv3892131 Chr.12:7721803 - 7835165 on Build GRCh38 Gain SLC2A14 CLEC4C NANOGNB NANOG
          dgv179n27 Chr.12:7818125 - 7946706 on Build GRCh38 Gain SLC2A14 NANOGP1 SLC2A3
          dgv61n111 Chr.12:7811433 - 7978362 on Build GRCh38 Duplication SLC2A14 NANOGP1 SLC2A3
          dgv2328n54 Chr.12:7800493 - 7935631 on Build GRCh38 Gain SLC2A14 NANOGP1 SLC2A3
          dgv1351n100 Chr.12:7801358 - 7927734 on Build GRCh38 Gain SLC2A14 NANOGP1 SLC2A3
          esv2745493 Chr.12:7778330 - 7906182 on Build GRCh38 Deletion SLC2A14 NANOGP1 NANOG
          esv3580494 Chr.12:7799167 - 7908106 on Build GRCh38 Gain SLC2A14 NANOGP1
          nsv1159953 Chr.12:7792788 - 7853910 on Build GRCh38 Duplication SLC2A14 NANOG
          dgv6e196 Chr.12:7670828 - 7920436 on Build GRCh38 Duplication SLC2A14 NANOGP1 GDF3 CLEC4C NANOGNB DPPA3 NANOG SLC2A3
          dgv1352n100 Chr.12:7805181 - 7978362 on Build GRCh38 Gain SLC2A14 NANOGP1 SLC2A3
          dgv57e55 Chr.12:7700916 - 7963472 on Build GRCh38 Gain SLC2A14 NANOGP1 CLEC4C NANOGNB DPPA3 NANOG SLC2A3
          dgv5e196 Chr.12:7621924 - 7954221 on Build GRCh38 Duplication SLC2A14 NANOGP1 GDF3 APOBEC1 CLEC4C NANOGNB DPPA3 NANOG SLC2A3
          nsv983268 Chr.12:7810216 - 7821299 on Build GRCh38 Duplication SLC2A14

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          cell differentiation
          glucose transmembrane transport
          glucose transmembrane transporter activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline