Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03198166_cn
          See other CCL3L3 CNV Assays ›
          Gene Symbol
          CCL3L3
          Assay Reference Genome
          Location

          Chr.17:36196733 on build GRCh38
          Cytoband
          17q12
          Assay ID Hs03198166_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          414062

          Gene Name:

          C-C motif chemokine ligand 3 like 3

          Gene Aliases:

          464.2, D17S1718, G0S19-2, LD78, LD78BETA, SCYA3L, SCYA3L1

          Location:

          Chr.17:36194869-36196748 on Build GRCh38

          Assay Gene Location:

          Overlaps - Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CCL3L3 NM_001001437.3 NP_001001437.2
          AA380745.1
          BC007783.2 AAH07783.1
          BC063472.1
          BC146919.1
          BC147008.1
          BC147013.1
          CF128666.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv514837 Chr.17:36175569 - 36196796 on Build GRCh38 Gain CCL3L3 TBC1D3B
          esv3333423 Chr.17:36196240 - 36197637 on Build GRCh38 Duplication CCL3L3
          dgv5541n54 Chr.17:36180353 - 36217165 on Build GRCh38 Loss CCL3L3 CCL4L2
          nsv471397 Chr.17:36194872 - 36196757 on Build GRCh38 Gain CCL3L3
          nsv820772 Chr.17:36176743 - 36222726 on Build GRCh38 Deletion CCL3L3 CCL4L2
          dgv5540n54 Chr.17:36179341 - 36265379 on Build GRCh38 Loss CCL3L3 CCL4L2 TBC1D3I
          dgv3147n100 Chr.17:36102943 - 36215877 on Build GRCh38 Gain CCL3L3 CCL4L2 CCL4 TBC1D3B
          nsv1143852 Chr.17:36180924 - 36242254 on Build GRCh38 Deletion CCL3L3 CCL4L2
          nsv1133405 Chr.17:36177240 - 36223356 on Build GRCh38 Deletion CCL3L3 CCL4L2
          nsv1066186 Chr.17:36132993 - 36208237 on Build GRCh38 Gain CCL3L3 TBC1D3B
          dgv3145n100 Chr.17:36097969 - 36258784 on Build GRCh38 Gain CCL3L3 CCL4L2 CCL4 TBC1D3I TBC1D3B
          nsv960473 Chr.17:36154369 - 36242140 on Build GRCh38 Duplication CCL3L3 CCL4L2 TBC1D3B
          dgv5538n54 Chr.17:36118154 - 36215910 on Build GRCh38 Gain CCL3L3 CCL4L2 TBC1D3B
          esv3374119 Chr.17:36190073 - 36197466 on Build GRCh38 Duplication CCL3L3
          nsv1160469 Chr.17:36109315 - 36200725 on Build GRCh38 Duplication CCL3L3 TBC1D3B
          dgv5536n54 Chr.17:36107306 - 36198514 on Build GRCh38 Gain CCL3L3 TBC1D3B
          esv2760467 Chr.17:36108119 - 36265379 on Build GRCh38 Gain CCL3L3 CCL4L2 TBC1D3I TBC1D3B
          dgv3146n100 Chr.17:36098851 - 36208237 on Build GRCh38 Gain CCL3L3 CCL4 TBC1D3B
          esv3893005 Chr.17:36153146 - 36215910 on Build GRCh38 Loss CCL3L3 CCL4L2 TBC1D3B
          nsv2040 Chr.17:36147099 - 36265379 on Build GRCh38 Deletion CCL3L3 CCL4L2 TBC1D3I TBC1D3B
          nsv1072731 Chr.17:36181223 - 36223656 on Build GRCh38 Deletion CCL3L3 CCL4L2
          dgv545e199 Chr.17:36181069 - 36227348 on Build GRCh38 Deletion CCL3L3 CCL4L2
          esv2762388 Chr.17:36060272 - 36265379 on Build GRCh38 Gain+Loss CCL18 CCL3L3 CCL4L2 CCL4 TBC1D3I CCL3 TBC1D3B
          nsv574781 Chr.17:36181006 - 36214601 on Build GRCh38 Gain CCL3L3 CCL4L2
          esv33107 Chr.17:36105177 - 36265379 on Build GRCh38 Loss CCL3L3 CCL4L2 CCL4 TBC1D3I TBC1D3B
          dgv3144n100 Chr.17:36089191 - 36265379 on Build GRCh38 Gain CCL3L3 CCL4L2 CCL4 TBC1D3I CCL3 TBC1D3B
          nsv1061066 Chr.17:36084283 - 36258784 on Build GRCh38 Loss CCL3L3 CCL4L2 CCL4 TBC1D3I CCL3 TBC1D3B
          esv2715876 Chr.17:36098855 - 36206226 on Build GRCh38 Deletion CCL3L3 CCL4 TBC1D3B
          nsv433448 Chr.17:36110820 - 36215910 on Build GRCh38 Gain CCL3L3 CCL4L2 TBC1D3B
          nsv953882 Chr.17:36182421 - 36224156 on Build GRCh38 Deletion CCL3L3 CCL4L2

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation Validated

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          monocyte chemotaxis
          inflammatory response
          G-protein coupled receptor signaling pathway
          negative regulation of cell proliferation
          neutrophil chemotaxis
          positive regulation of GTPase activity
          lymphocyte chemotaxis
          positive regulation of inflammatory response
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          chemokine activity
          CCR chemokine receptor binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0