Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03378820_cn
          See other CHD1L CNV Assays ›
          Gene Symbol
          CHD1L
          Assay Reference Genome
          Location

          Chr.1:147183845 on build GRCh38
          Cytoband
          1q21.1
          Assay ID Hs03378820_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9557

          Gene Name:

          chromodomain helicase DNA binding protein 1 like

          Gene Aliases:

          ALC1, CHDL

          Location:

          Chr.1:147172804-147295766 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CHD1L XM_017002858.1 XP_016858347.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain LINC01138 LINC00624 LOC105371242 NUDT4P2 PPIAL4G PDIA3P1 RNU1-13P MIR6077 MIR5087 GJA8 RNVU1-7 FMO5 BCL9 CHD1L LOC101928979 RNVU1-3 LOC101927468 LOC728989 RNVU1-8 NBPF14 HYDIN2 RNVU1-1 LOC100996517 NUDT4P1 LOC100996763 NBPF11 NBPF10 GJA5 LOC100505824 LOC100132057 NBPF13P NOTCH2NL GPR89B PRKAB2 LOC284561 LOC105371226 PDZK1P1 NBPF12 RNVU1-6 ACP6 PDE4DIP CH17-408M7.1
          nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss CD160 HFE2 RNF115 PPIAL4G MIR6077 GJA8 NUDT17 RNVU1-3 LOC728989 NBPF14 GNRHR2 RNVU1-1 LOC100996517 NUDT4P1 NBPF10 GJA5 LIX1L PRKAB2 LOC284561 LOC105371226 PDZK1P1 RBM8A ACP6 PDE4DIP CH17-408M7.1 LINC01138 LINC00624 LOC105371242 NUDT4P2 PDIA3P1 RNU1-13P POLR3GL MIR5087 ITGA10 RNVU1-7 PDZK1 FMO5 BCL9 CHD1L ANKRD35 POLR3C MIR6736 LOC101928979 LOC101927468 RNVU1-8 TXNIP HYDIN2 LOC100996763 NBPF11 LOC100505824 LOC100132057 GPR89A NBPF13P NOTCH2NL GPR89B NBPF12 RNVU1-6 PIAS3 PEX11B ANKRD34A
          nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion NBPF13P LOC728989 FMO5 RNVU1-8 PRKAB2 CHD1L PDIA3P1
          esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss LINC01138 LINC00624 LOC105371242 NUDT4P2 PPIAL4G PDIA3P1 RNU1-13P MIR6077 MIR5087 GJA8 RNVU1-7 FMO5 BCL9 CHD1L RNVU1-3 LOC101927468 LOC728989 RNVU1-8 NBPF14 HYDIN2 RNVU1-1 LOC100996517 NUDT4P1 LOC100996763 NBPF11 NBPF10 GJA5 LOC100505824 LOC100132057 NBPF13P NOTCH2NL GPR89B PRKAB2 LOC284561 LOC105371226 PDZK1P1 NBPF12 ACP6 PDE4DIP CH17-408M7.1
          nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion LINC01138 LINC00624 HFE2 LOC105371242 NUDT4P2 PPIAL4G PDIA3P1 RNU1-13P POLR3GL MIR6077 MIR5087 GJA8 RNVU1-7 FMO5 BCL9 CHD1L LOC101928979 RNVU1-3 LOC101927468 LOC728989 RNVU1-8 NBPF14 TXNIP HYDIN2 RNVU1-1 LOC100996517 NUDT4P1 LOC100996763 NBPF11 NBPF10 GJA5 LOC100505824 LOC100132057 NBPF13P LIX1L NOTCH2NL GPR89B PRKAB2 LOC284561 LOC105371226 PDZK1P1 NBPF12 RNVU1-6 ACP6 PDE4DIP ANKRD34A CH17-408M7.1
          nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication LINC01138 LINC00624 LOC105371242 NUDT4P2 PPIAL4G PDIA3P1 RNU1-13P MIR6077 MIR5087 GJA8 RNVU1-7 FMO5 BCL9 CHD1L RNVU1-3 LOC101927468 LOC728989 RNVU1-8 NBPF14 HYDIN2 RNVU1-1 LOC100996517 NUDT4P1 LOC100996763 NBPF11 GJA5 LOC100505824 LOC100132057 NBPF13P NOTCH2NL GPR89B PRKAB2 LOC284561 LOC105371226 PDZK1P1 NBPF12 ACP6 PDE4DIP CH17-408M7.1
          nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain LINC01138 LINC00624 LOC105371242 NUDT4P2 PPIAL4G PDIA3P1 RNU1-13P MIR6077 MIR5087 GJA8 RNVU1-7 FMO5 BCL9 CHD1L LOC101928979 RNVU1-3 LOC101927468 LOC728989 RNVU1-8 NBPF14 HYDIN2 RNVU1-1 LOC100996517 NUDT4P1 LOC100996763 NBPF11 NBPF10 GJA5 LOC100505824 LOC100132057 NBPF13P NOTCH2NL GPR89B PRKAB2 LOC284561 LOC105371226 PDZK1P1 NBPF12 RNVU1-6 ACP6 PDE4DIP CH17-408M7.1
          dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion LINC01138 LINC00624 LOC105371242 NUDT4P2 PPIAL4G PDIA3P1 RNU1-13P MIR6077 MIR5087 GJA8 RNVU1-7 FMO5 BCL9 CHD1L NBPF9 LOC101928979 RNVU1-3 LOC101927468 LOC728989 RNVU1-8 NBPF14 HYDIN2 RNVU1-1 LOC100996517 NUDT4P1 LOC100996763 NBPF11 NBPF10 GJA5 LOC100505824 LOC100132057 NBPF13P NOTCH2NL GPR89B PRKAB2 LOC284561 LOC105371226 PDZK1P1 NBPF12 ACP6 PDE4DIP CH17-408M7.1
          nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss FMO5 PRKAB2 CHD1L PDIA3P1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair, DNA duplex unwinding
          DNA repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          chromatin remodeling
          cellular response to DNA damage stimulus
          nucleotide-excision repair, DNA incision
          global genome nucleotide-excision repair
          nucleotide binding
          nucleic acid binding
          ATP-dependent DNA helicase activity
          protein binding
          ATP binding
          ATPase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0