Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03618933_cn
          See other HLA-DPA1 CNV Assays ›
          Gene Symbol
          HLA-DPA1
          Assay Reference Genome
          Location

          Chr.6:33075989 on build GRCh38
          Cytoband
          6p21.32
          Assay ID Hs03618933_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3113

          Gene Name:

          major histocompatibility complex, class II, DP alpha 1

          Gene Aliases:

          DP(W3), DP(W4), HLA-DP1A, HLADP, HLASB, PLT1

          Location:

          Chr.6:33064569-33080778 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          HLA-DPA1 NM_001242524.1 NP_001229453.1
          NM_001242525.1 NP_001229454.1
          BI553587.1
          CD516536.1

          Target Gene Details

          Entrez Gene ID:

          3115

          Gene Name:

          major histocompatibility complex, class II, DP beta 1

          Gene Aliases:

          DPB1, HLA-DP, HLA-DP1B, HLA-DPB

          Location:

          Chr.6:33075926-33089696 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          HLA-DPB1 NM_002121.5 1 64 NP_002112.3
          AB208978.1 BAD92215.1
          AK291986.1
          AK301322.1
          AK303153.1
          AK313305.1
          AY656678.1 AAV71025.1
          AY656679.1 AAV71026.1
          AY656680.1 AAV71027.1
          AY804132.1 AAW78737.1
          AY804133.1 AAW78738.1
          AY804134.1 AAW78739.1
          AY804135.1 AAW78740.1
          AY804136.1 AAW78741.1
          AY804137.1 AAW78742.1
          AY804138.1 AAW78743.1
          AY804139.1 AAW78744.1
          AY804140.1 AAW78745.1
          AY804141.1 AAW78746.1
          AY804142.1 AAW78747.1
          AY804143.1 AAW78748.1
          AY831401.1 AAW80919.1
          AY831402.1 AAW80920.1
          AY831403.1 AAW80921.1
          AY831404.1 AAW80922.1
          BC007963.2 AAH07963.1
          BC013184.1 AAH13184.1
          BC015000.1 AAH15000.1
          BP213121.1 1 64
          BT019780.1 AAV38583.1
          DQ284441.1 ABC66207.1
          FR775797.1
          GQ891549.1
          HE603611.1
          HE603612.1
          HE614584.1
          HE614585.1
          HE614586.1
          HE614587.1
          HE614588.1
          HE614589.1
          HE614590.1
          HE614591.1
          J03041.1 AAA35993.1
          KJ780721.1
          M28200.1 AAA53195.1
          M57466.1 AAA63221.1
          M83664.1 AAA59837.1
          X01426.1 CAA25672.1
          X03067.1 CAA26871.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv823645 Chr.6:33061191 - 33091364 on Build GRCh38 Loss HLA-DPA1 HLA-DPB1
          esv2731879 Chr.6:33065144 - 33107309 on Build GRCh38 Deletion HLA-DPA1 HLA-DPB1
          nsv1024429 Chr.6:32997190 - 33154554 on Build GRCh38 Loss HLA-DPA1 HLA-DPB1 HLA-DOA HLA-DPB2 HCG24
          esv1006913 Chr.6:33064893 - 33088398 on Build GRCh38 Gain HLA-DPA1 HLA-DPB1
          nsv1073969 Chr.6:31456422 - 33480523 on Build GRCh38 Deletion FKBPL HLA-DQB2 PRRC2A HLA-DRA HLA-DPA1 ATP6V1G2 NCR3 CYP21A2 GPSM3 SNORA38 SNORD48 AGER APOM MICB SKIV2L ZBTB9 MIR219A1 HSD17B8 ZBTB12 RING1 MIR6873 AIF1 EGFL8 SYNGAP1 HLA-DMB RPS18 C6orf10 VARS HLA-DOA GPANK1 VPS52 SLC39A7 LY6G5C NELFE TNXA MIR5004 LST1 MSH5 NFKBIL1 MIR6721 PFDN6 B3GALT4 C6orf47 HLA-DQB1-AS1 LY6G6F ABHD16A SNORD117 STK19 LY6G6D RGL2 HLA-DOB HCG25 EHMT2 SAPCD1-AS1 HCG26 DDAH2 MIR6832 NEU1 HLA-DRB6 MIR1236 MIR4646 MIR6834 PPT2-EGFL8 C2 MIR3135B VWA7 DDX39B LOC100287329 COL11A2 HLA-DQA1 TNXB SNORD84 HLA-DQA2 DXO CSNK2B HSPA1B CUTA LY6G5B HCP5 HLA-DRB5 LOC100294145 BAG6 LY6G6E HCG23 HLA-DPB2 LTB PRRT1 HSPA1L TNF ATF6B PBX2 CFB HSPA1A BRD2 C4A HCG24 C6orf25 PSMB8 HLA-DRB1 LOC100507547 WDR46 KIFC1 LOC101929163 LTA CLIC1 SNORD52 ATP6V1G2-DDX39B MCCD1 PSMB8-AS1 TAP1 LOC102725068 DAXX PPP1R2P1 PPT2 BTNL2 AGPAT1 C6orf48 TAP2 RXRB MSH5-SAPCD1 DDX39B-AS1 NOTCH4 RNF5 PSMB9 HLA-DQB1 TAPBP SAPCD1 MIR6833 C2-AS1 PHF1 C4B ZBTB22 SLC44A4 HLA-DMA CYP21A1P LSM2 HLA-DPB1 LY6G6C
          nsv1112900 Chr.6:31455323 - 33480423 on Build GRCh38 Deletion FKBPL HLA-DQB2 PRRC2A HLA-DRA HLA-DPA1 ATP6V1G2 NCR3 CYP21A2 GPSM3 SNORA38 SNORD48 AGER APOM MICB SKIV2L ZBTB9 MIR219A1 HSD17B8 ZBTB12 RING1 MIR6873 AIF1 EGFL8 SYNGAP1 HLA-DMB RPS18 C6orf10 VARS HLA-DOA GPANK1 VPS52 SLC39A7 LY6G5C NELFE TNXA MIR5004 LST1 MSH5 NFKBIL1 MIR6721 PFDN6 B3GALT4 C6orf47 HLA-DQB1-AS1 LY6G6F ABHD16A SNORD117 STK19 LY6G6D RGL2 HLA-DOB HCG25 EHMT2 SAPCD1-AS1 HCG26 DDAH2 MIR6832 NEU1 HLA-DRB6 MIR1236 MIR4646 MIR6834 PPT2-EGFL8 C2 MIR3135B VWA7 DDX39B LOC100287329 COL11A2 HLA-DQA1 TNXB SNORD84 HLA-DQA2 DXO CSNK2B HSPA1B CUTA LY6G5B HCP5 HLA-DRB5 LOC100294145 BAG6 LY6G6E HCG23 HLA-DPB2 LTB PRRT1 HSPA1L TNF ATF6B PBX2 CFB HSPA1A BRD2 C4A HCG24 C6orf25 PSMB8 HLA-DRB1 LOC100507547 WDR46 KIFC1 LOC101929163 LTA CLIC1 SNORD52 ATP6V1G2-DDX39B MCCD1 PSMB8-AS1 TAP1 LOC102725068 DAXX PPP1R2P1 PPT2 BTNL2 AGPAT1 C6orf48 TAP2 RXRB MSH5-SAPCD1 DDX39B-AS1 NOTCH4 RNF5 PSMB9 HLA-DQB1 TAPBP SAPCD1 MIR6833 C2-AS1 PHF1 C4B ZBTB22 SLC44A4 HLA-DMA CYP21A1P LSM2 HLA-DPB1 LY6G6C
          esv22467 Chr.6:33055021 - 33087568 on Build GRCh38 Loss HLA-DPA1 HLA-DPB1
          nsv602818 Chr.6:32998165 - 33150041 on Build GRCh38 Loss HLA-DPA1 HLA-DPB1 HLA-DOA HLA-DPB2 HCG24
          nsv1119076 Chr.6:32595823 - 33480623 on Build GRCh38 Deletion WDR46 RPS18 KIFC1 MIR3135B HLA-DOA PSMB8-AS1 HLA-DQB2 TAP1 VPS52 SLC39A7 COL11A2 HLA-DQA1 DAXX PPP1R2P1 HLA-DPA1 MIR5004 TAP2 RXRB HLA-DQA2 PFDN6 B3GALT4 PSMB9 HLA-DQB1 CUTA TAPBP LOC100294145 HLA-DQB1-AS1 HLA-DPB2 PHF1 ZBTB9 RGL2 ZBTB22 HLA-DOB HCG25 MIR219A1 HSD17B8 HLA-DMA HLA-DPB1 BRD2 HCG24 RING1 MIR6873 SYNGAP1 MIR6834 PSMB8 HLA-DMB

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          positive regulation of interferon-gamma production
          positive regulation of T cell proliferation
          T cell receptor signaling pathway
          positive regulation of T cell activation
          interferon-gamma-mediated signaling pathway
          cellular response to interferon-gamma
          MHC class II receptor activity
          peptide antigen binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline