Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03723562_cn
          See other SIGMAR1 CNV Assays ›
          Gene Symbol
          SIGMAR1
          Assay Reference Genome
          Location

          Chr.9:34636736 on build GRCh38
          Cytoband
          Assay ID Hs03723562_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          10280

          Gene Name:

          sigma non-opioid intracellular receptor 1

          Gene Aliases:

          ALS16, DSMA2, HMNR2, OPRS1, SIG-1R, SR-BP, SR-BP1, SRBP, hSigmaR1, sigma1R

          Location:

          Chr.9:34634722-34637787 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SIGMAR1 NM_001282205.2 NP_001269134.1
          NM_001282206.2 NP_001269135.1
          NM_001282207.2 NP_001269136.1
          NM_001282208.2 NP_001269137.1
          NM_001282209.2 NP_001269138.1
          NM_005866.4 NP_005857.1
          NM_147157.3 NP_671513.1
          NR_104108.2
          AF226604.1 AAF64280.1
          AK098451.1 BAC05307.1
          AK130502.1
          AK222899.1 BAD96619.1
          AK291092.1 BAF83781.1
          AK299130.1 BAG61183.1
          AY633611.1 AAV33304.1
          BC004899.2 AAH04899.1
          BC007839.2 AAH07839.1
          BI596428.1
          CR457075.1 CAG33356.1
          DQ644568.1 ABG29111.1
          DQ647702.1 ABG36559.1
          DQ656583.1 ABG46369.1
          U75283.1 AAB50402.1
          U79528.1 AAB51238.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3620335 Chr.9:33871811 - 34738198 on Build GRCh38 Gain LOC114224 NUDT2 SNORD121B DCTN3 RPP25L SNORD121A PHF24 CCL21 SIGMAR1 FAM219A CNTFR ENHO UBAP1 KIF24 UBE2R2 ARID3C CCL19 SPMIP6 CNTFR-AS1 MYORG LOC730098 IL11RA UBAP2 DNAI1 DCAF12 CCL27 GALT SPATA31F1
          nsv6449537 Chr.9:34103197 - 34805619 on Build GRCh38 Duplication LOC114224 NUDT2 ARID3C DCTN3 CCL19 SPMIP6 RPP25L CNTFR-AS1 MYORG LOC730098 IL11RA PHF24 CCL21 SIGMAR1 FAM219A DNAI1 CNTFR DCAF12 CCL27 ENHO UBAP1 GALT KIF24 SPATA31F1
          nsv1161857 Chr.9:34537279 - 34669174 on Build GRCh38 Deletion SIGMAR1 ARID3C CNTFR DCTN3 CCL27 GALT RPP25L CNTFR-AS1 LOC730098 IL11RA PHF24
          dgv12659n54 Chr.9:34552439 - 34663141 on Build GRCh38 Loss SIGMAR1 ARID3C CNTFR DCTN3 CCL27 GALT RPP25L CNTFR-AS1 IL11RA
          nsv4173877 Chr.9:34598462 - 34640256 on Build GRCh38 Deletion SIGMAR1 ARID3C DCTN3 RPP25L
          nsv818699 Chr.9:34535211 - 34685943 on Build GRCh38 Gain SIGMAR1 ARID3C CNTFR DCTN3 CCL27 GALT RPP25L CNTFR-AS1 LOC730098 IL11RA PHF24

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid transport
          nervous system development
          opioid receptor signaling pathway
          regulation of neuron apoptotic process
          protein homotrimerization
          regulation of postsynapse assembly
          opioid receptor activity
          protein binding
          signaling receptor activity
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline