Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03837016_cn
          See other ZNF10 CNV Assays ›
          Gene Symbol
          ZNF10
          Assay Reference Genome
          Location

          Chr.12:133133504 on build GRCh38
          Cytoband
          12q24.33
          Assay ID Hs03837016_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          7556

          Gene Name:

          zinc finger protein 10

          Gene Aliases:

          KOX1

          Location:

          Chr.12:133130628-133159465 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ZNF10 NM_015394.4 NP_056209.2
          AK314784.1
          BC024182.2 AAH24182.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3631327 Chr.12:133059578 - 133181486 on Build GRCh38 Gain ZNF268 ZNF10 ZNF84 ZNF891 ZNF140
          dgv1595n100 Chr.12:133097005 - 133183162 on Build GRCh38 Gain ZNF268 ZNF10 ZNF891 ZNF140
          nsv470337 Chr.12:133100763 - 133202210 on Build GRCh38 Loss ZNF268 ZNF10 ZNF891 ZNF140
          dgv1593n100 Chr.12:133062687 - 133201059 on Build GRCh38 Gain ZNF268 ZNF10 ZNF84 ZNF891 ZNF140
          dgv1596n100 Chr.12:133121951 - 133201059 on Build GRCh38 Gain ZNF268 ZNF10 ZNF891
          dgv3060n54 Chr.12:133104113 - 133202210 on Build GRCh38 Gain ZNF268 ZNF10 ZNF891 ZNF140
          dgv1594n100 Chr.12:133084713 - 133150038 on Build GRCh38 Gain ZNF10 ZNF891 ZNF140
          esv27162 Chr.12:133060920 - 133202839 on Build GRCh38 Gain+Loss ZNF268 ZNF10 ZNF84 ZNF891 ZNF140
          dgv534e212 Chr.12:133105970 - 133183134 on Build GRCh38 Gain ZNF268 ZNF10 ZNF891 ZNF140
          nsv1054203 Chr.12:133115480 - 133160725 on Build GRCh38 Gain ZNF10 ZNF891
          nsv455818 Chr.12:133127915 - 133202210 on Build GRCh38 Loss ZNF268 ZNF10 ZNF891
          esv2763014 Chr.12:133115492 - 133201603 on Build GRCh38 Gain ZNF268 ZNF10 ZNF891
          dgv1590n100 Chr.12:132973304 - 133201059 on Build GRCh38 Gain ZNF26 LOC101928597 ZNF268 ZNF10 ZNF84 ZNF891 ZNF140
          esv2760921 Chr.12:133076187 - 133201603 on Build GRCh38 Gain ZNF268 ZNF10 ZNF891 ZNF140
          esv2759923 Chr.12:132908635 - 133157527 on Build GRCh38 Loss ZNF26 LOC101928597 ZNF10 ZNF84 ZNF891 ZNF140 ZNF605
          dgv220n27 Chr.12:133107735 - 133202210 on Build GRCh38 Gain ZNF268 ZNF10 ZNF891

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0