Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03838785_cn
          See other SLC2A14 CNV Assays ›
          Gene Symbol
          SLC2A14
          Assay Reference Genome
          Location

          Chr.12:7873924 on build GRCh38
          Cytoband
          12p13.31
          Assay ID Hs03838785_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          144195

          Gene Name:

          solute carrier family 2 member 14

          Gene Aliases:

          GLUT14, SLC2A3P3

          Location:

          Chr.12:7812512-7891196 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SLC2A14 NM_001286237.1 NP_001273166.1
          XM_011520565.2 XP_011518867.1
          XM_017018847.1 XP_016874336.1
          AK299037.1
          AK316238.1
          DA723309.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2656717 Chr.12:7798646 - 7905603 on Build GRCh38 Deletion SLC2A14 NANOGP1
          dgv2334n54 Chr.12:7847740 - 7950607 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          esv2751114 Chr.12:7810616 - 7964473 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          esv3580097 Chr.12:7851816 - 7971452 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          nsv826225 Chr.12:7845109 - 7974566 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv59e55 Chr.12:7864877 - 7989987 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          nsv1035735 Chr.12:7851817 - 7882543 on Build GRCh38 Gain SLC2A14
          esv29743 Chr.12:7842682 - 7970710 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv61n111 Chr.12:7811433 - 7978362 on Build GRCh38 Duplication SLC2A3 SLC2A14 NANOGP1
          esv2745493 Chr.12:7778330 - 7906182 on Build GRCh38 Deletion SLC2A14 NANOGP1 NANOG
          nsv469100 Chr.12:7871766 - 7946706 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv6e196 Chr.12:7670828 - 7920436 on Build GRCh38 Duplication SLC2A3 SLC2A14 CLEC4C NANOGP1 NANOGNB DPPA3 GDF3 NANOG
          nsv470265 Chr.12:7840720 - 7970710 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          nsv818883 Chr.12:7832345 - 7916285 on Build GRCh38 Gain SLC2A14 NANOGP1
          dgv2335n54 Chr.12:7848316 - 7973149 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          esv3548706 Chr.12:7842215 - 7980904 on Build GRCh38 Deletion SLC2A3 SLC2A14 NANOGP1
          esv3628445 Chr.12:7847526 - 8064130 on Build GRCh38 Gain SLC2A3 C3AR1 SLC2A14 FOXJ2 NANOGP1
          dgv58e55 Chr.12:7801455 - 7987805 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv2336n54 Chr.12:7869336 - 7938243 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          esv2759877 Chr.12:7704223 - 8471391 on Build GRCh38 Gain+Loss SLC2A3 SLC2A14 CLEC6A CLEC4C FAM86FP FAM66C DPPA3 NECAP1 ZNF705A NANOG C3AR1 POU5F1P3 FOXJ2 CLEC4A NANOGP1 NANOGNB FAM90A1 LOC101927905 LINC00937
          dgv2329n54 Chr.12:7808980 - 7993566 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          nsv1070090 Chr.12:7798719 - 7905639 on Build GRCh38 Deletion SLC2A14 NANOGP1
          dgv60e55 Chr.12:7864882 - 7982073 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          dgv11e213 Chr.12:7851829 - 7987805 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv1354n100 Chr.12:7842179 - 7952475 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv166e201 Chr.12:7827227 - 7939801 on Build GRCh38 Deletion SLC2A3 SLC2A14 NANOGP1
          dgv179n27 Chr.12:7818125 - 7946706 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv2328n54 Chr.12:7800493 - 7935631 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv1351n100 Chr.12:7801358 - 7927734 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          esv2760250 Chr.12:7842191 - 7978362 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv167e201 Chr.12:7842493 - 7972788 on Build GRCh38 Deletion SLC2A3 SLC2A14 NANOGP1
          esv3580494 Chr.12:7799167 - 7908106 on Build GRCh38 Gain SLC2A14 NANOGP1
          nsv975644 Chr.12:7843169 - 7915838 on Build GRCh38 Duplication SLC2A14 NANOGP1
          dgv449e212 Chr.12:7821394 - 7989987 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          dgv1352n100 Chr.12:7805181 - 7978362 on Build GRCh38 Gain SLC2A3 SLC2A14 NANOGP1
          esv3892130 Chr.12:7824755 - 7923089 on Build GRCh38 Gain+Loss SLC2A3 SLC2A14 NANOGP1
          dgv183n27 Chr.12:7844294 - 7973149 on Build GRCh38 Loss SLC2A3 SLC2A14 NANOGP1
          dgv57e55 Chr.12:7700916 - 7963472 on Build GRCh38 Gain SLC2A3 SLC2A14 CLEC4C NANOGP1 NANOGNB DPPA3 NANOG
          dgv5e196 Chr.12:7621924 - 7954221 on Build GRCh38 Duplication SLC2A3 SLC2A14 CLEC4C NANOGP1 NANOGNB APOBEC1 DPPA3 GDF3 NANOG

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          cell differentiation
          glucose transmembrane transport
          glucose transmembrane transporter activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline