Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03860935_cn
          See other SGCG CNV Assays ›
          Gene Symbol
          SGCG
          Assay Reference Genome
          Location

          Chr.13:23168632 on build GRCh38
          Cytoband
          13q12.12
          Assay ID Hs03860935_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6445

          Gene Name:

          sarcoglycan gamma

          Gene Aliases:

          35DAG, A4, DAGA4, DMDA, DMDA1, LGMD2C, MAM, SCARMD2, SCG3, gamma-SG

          Location:

          Chr.13:23160308-23325165 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SGCG XM_006719861.3 XP_006719924.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2751135 Chr.13:22932509 - 23201361 on Build GRCh38 Gain SGCG
          nsv561292 Chr.13:23025847 - 24348376 on Build GRCh38 Loss TNFRSF19 SGCG NUS1P3 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 MIR2276
          nsv455838 Chr.13:22954546 - 23290518 on Build GRCh38 Gain SGCG
          esv2422437 Chr.13:22198584 - 23416312 on Build GRCh38 Duplication LINC00540 SGCG LINC00621 BASP1P1 SACS LOC105370109
          esv3892311 Chr.13:22674660 - 24411768 on Build GRCh38 Gain TNFRSF19 SGCG NUS1P3 LINC00621 BASP1P1 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 LOC105370109 MIR2276
          nsv1046767 Chr.13:22757080 - 23416839 on Build GRCh38 Gain SGCG LINC00621 BASP1P1 SACS LOC105370109
          esv3580576 Chr.13:22945778 - 24362710 on Build GRCh38 Loss TNFRSF19 SGCG NUS1P3 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 MIR2276
          nsv1040787 Chr.13:22929958 - 23294371 on Build GRCh38 Gain SGCG
          esv2422502 Chr.13:22755560 - 24382791 on Build GRCh38 Duplication TNFRSF19 SGCG NUS1P3 LINC00621 BASP1P1 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 LOC105370109 MIR2276
          nsv525965 Chr.13:22937442 - 24358507 on Build GRCh38 Gain TNFRSF19 SGCG NUS1P3 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 MIR2276
          dgv3084n54 Chr.13:22925582 - 24378278 on Build GRCh38 Gain TNFRSF19 SGCG NUS1P3 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 MIR2276
          nsv520725 Chr.13:23157247 - 23343142 on Build GRCh38 Loss SGCG SACS
          nsv1035266 Chr.13:23163833 - 23180307 on Build GRCh38 Gain SGCG
          nsv1046471 Chr.13:23095243 - 23316065 on Build GRCh38 Gain SGCG
          dgv1615n100 Chr.13:22930769 - 24362849 on Build GRCh38 Gain TNFRSF19 SGCG NUS1P3 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 MIR2276
          esv3631513 Chr.13:23146035 - 23217263 on Build GRCh38 Gain SGCG
          esv2751136 Chr.13:22982261 - 24393377 on Build GRCh38 Loss TNFRSF19 SGCG NUS1P3 SPATA13 SACS SACS-AS1 C1QTNF9B C1QTNF9B-AS1 LINC00566 MIPEP LINC00327 SPATA13-AS1 ANKRD20A19P C1QTNF9 MIR2276

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle organ development
          cardiac muscle tissue development
          muscle cell development
          heart contraction
          membrane organization
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0