Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03939200_cn
          See other CES1 CNV Assays ›
          Gene Symbol
          CES1
          Assay Reference Genome
          Location

          Chr.16:55831053 on build GRCh38
          Cytoband
          16q12.2
          Assay ID Hs03939200_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1066

          Gene Name:

          carboxylesterase 1

          Gene Aliases:

          ACAT, CE-1, CEH, CES2, HMSE, HMSE1, PCE-1, REH, SES1, TGH, hCE-1

          Location:

          Chr.16:55802851-55833186 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CES1 NM_001025194.1 NP_001020365.1
          NM_001025195.1 NP_001020366.1
          NM_001266.4 NP_001257.4
          XM_005255774.1 XP_005255831.1
          AB025026.1 BAA84995.1
          AB119995.1 BAC87748.1
          AB119996.1 BAC87749.1
          AF177775.1 AAD53175.1
          AK290623.1
          AK292209.1
          AK301651.1
          AU139632.1
          AY268104.1 AAP20868.1
          BC012418.1 AAH12418.1
          BC110338.1 AAI10339.1
          CB137639.1
          L07764.1 AAA16036.1
          L07765.1 AAA35711.1
          M73499.1 AAA35649.1
          S73751.1 AAC60631.2

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv5137n54 Chr.16:55808492 - 55835816 on Build GRCh38 Gain CES1
          esv32573 Chr.16:55809091 - 55831223 on Build GRCh38 Gain+Loss CES1
          dgv479n67 Chr.16:55808461 - 55832982 on Build GRCh38 Gain CES1
          dgv2987n100 Chr.16:55782722 - 55845205 on Build GRCh38 Gain CES1
          dgv840e212 Chr.16:55790795 - 55834514 on Build GRCh38 Gain CES1
          esv28619 Chr.16:55808213 - 55831467 on Build GRCh38 Gain CES1
          nsv820205 Chr.16:55793542 - 55832100 on Build GRCh38 Gain CES1
          dgv842e212 Chr.16:55806374 - 55834514 on Build GRCh38 Gain CES1
          nsv827676 Chr.16:55821542 - 55831399 on Build GRCh38 Gain CES1
          esv2758649 Chr.16:55635484 - 55949678 on Build GRCh38 Gain CES1P1 SLC6A2 CES1 CES5A CES1P2
          nsv572693 Chr.16:55809206 - 55844051 on Build GRCh38 Loss CES1
          dgv52e203 Chr.16:55750518 - 55837223 on Build GRCh38 Gain+Loss CES1P1 CES1
          nsv9446 Chr.16:55806461 - 55833345 on Build GRCh38 Gain CES1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          metabolic process
          response to toxic substance
          epithelial cell differentiation
          methylumbelliferyl-acetate deacetylase activity
          carboxylic ester hydrolase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lvxnv:80/100.66.77.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0