Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03981389_cn
          See other DUSP14 CNV Assays ›
          Gene Symbol
          DUSP14
          Assay Reference Genome
          Location

          Chr.17:1730160 on build GRCh38
          Cytoband
          Assay ID Hs03981389_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          11072

          Gene Name:

          dual specificity phosphatase 14

          Gene Aliases:

          MKP-L, MKP6

          Location:

          Chr.17:1725025-1752665 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          DUSP14

          Target Gene Details

          Entrez Gene ID:

          4905

          Gene Name:

          N-ethylmaleimide sensitive factor, vesicle fusing ATPase

          Gene Aliases:

          DEE96, SEC18, SKD2

          Location:

          Chr.17:1110346-1276876 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NSF

          Target Gene Details

          Entrez Gene ID:

          124997

          Gene Name:

          WD repeat domain 81

          Gene Aliases:

          CAMRQ2, CHMRQ, HYC3, PPP1R166, SORF-2

          Location:

          Chr.17:146551-168624 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          WDR81 NM_001163673.2 NP_001157145.1
          NM_001163809.2 NP_001157281.1
          NM_001163811.2 NP_001157283.1
          NM_152348.4 NP_689561.2
          XM_011523651.3 XP_011521953.1
          XM_017024184.2 XP_016879673.1
          XM_047435340.1 XP_047291296.1
          XM_047435341.1 XP_047291297.1
          AK074111.1 BAB84937.1
          AK091136.1 BAC03593.1
          AK122972.1
          AK123896.1 BAG53978.1
          AK127946.1 BAG54603.1
          AK298567.1 BAH12815.1
          AL834379.1 CAD39042.1
          BC092513.1 AAH92513.1
          BC114519.1 AAI14520.1
          BC114568.1 AAI14569.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv9490 Chr.17:1653428 - 1828993 on Build GRCh38 Loss TADA2A SMYD4 SERPINF1 SERPINF2 MIR22 PRPF8 NSF MIR22HG WDR81 SYNRG WNT3 DUSP14 TLCD2
          nsv4237808 Chr.17:1582120 - 2528622 on Build GRCh38 Duplication TADA2A SMYD4 MIR22 DPH1 TSR1 MIR378J METTL16 SYNRG ACACA GPR179 SOCS7 C17orf78 SGSM2 TBC1D3E HIC1 PRPF8 LOC101927864 MIR22HG SMG6-AS1 WNT3 DDX52 SRR MNT SERPINF2 ARHGAP23 RTN4RL1 RILP TBC1D3 NPEPPSP1 NSF MRPL45 SCARF1 DUSP14 TLCD2 SNORD91B MIR132 SMG6 YWHAEP7 HNF1B OVCA2 SERPINF1 LOC284009 RPA1 LOC105371756 LOC101927839 SNORD91A MIR212 WDR81 RTN4RL1-AS1 SLC43A2
          nsv516886 Chr.17:1708905 - 1754359 on Build GRCh38 Loss TADA2A NSF MIR22HG WDR81 SERPINF2 MIR22 SYNRG WNT3 DUSP14 TLCD2
          nsv1946 Chr.17:1717490 - 1781570 on Build GRCh38 Insertion TADA2A SMYD4 NSF SERPINF1 WDR81 SERPINF2 SYNRG WNT3 DUSP14
          nsv833335 Chr.17:1713955 - 1827844 on Build GRCh38 Gain+Loss TADA2A SMYD4 NSF MIR22HG SERPINF1 WDR81 SERPINF2 MIR22 SYNRG WNT3 DUSP14
          nsv6511128 Chr.17:1727705 - 1921432 on Build GRCh38 Duplication SMYD4 MIR378J NSF SERPINF1 WDR81 RPA1 SERPINF2 SYNRG WNT3 DUSP14 LOC105371756 DDX52

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          mitochondrion organization
          aggrephagy
          early endosome to late endosome transport
          protein stabilization
          protein binding
          phosphatidylinositol 3-kinase regulator activity
          K63-linked polyubiquitin binding
          phosphatidylinositol 3-kinase inhibitor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline