Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04011945_cn
          Assay Reference Genome
          Location

          Chr.19:49512495 on build GRCh38
          Cytoband
          Assay ID Hs04011945_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv9739 Chr.19:47901840 - 50143063 on Build GRCh38 Gain+Loss NOSIP SNAR-A12 DKKL1 GRWD1 PNKP DHDH IRF3 SNORD32A PTOV1-AS1 CYTH2 BAX MIR4750 RRAS SNAR-A10 CGB2 SNAR-A13 DBP PPFIA3 MIR5088 MIR6799 BCL2L12 SYNGR4 CD37 SNAR-A9 SNAR-D SNAR-A14 SNAR-A5 FUT2 PLA2G4C-AS1 SNAR-A8 C19orf73 RPS11 SPHK2 ZNF114 CPT1C RUVBL2 CGB7 SNAR-G2 CGB3 SNAR-B2 SNAR-A7 LIG1 NTN5 MIR4751 NUCB1-AS1 CA11 AKT1S1 SNAR-C5 PRMT1 ALDH16A1 FUZ SCAF1 PRR12 RCN3 LOC105372435 TSKS SIGLEC16 TEAD2 LOC105372430 BSPH1 KCNJ14 HSD17B14 SNAR-G1 SNAR-C1 FGF21 EMP3 PLA2G4C MAMSTR CABP5 SLC17A7 LMTK3 NUP62 LOC101928295 SULT2B1 CGB8 PLEKHA4 TMEM143 SNAR-B1 IL4I1 PRRG2 RPL13A TULP2 FTL SNAR-A6 FLT3LG SNORD35A MIR6798 FAM83E KDELR1 SNORD34 PPP1R15A ZNF473 NTF4 CGB1 VRK3 RASIP1 SEC1P SPACA4 MIR6800 LHB NUCB1 SNAR-C3 CGB5 TBC1D17 SLC6A16 CARD8 MIR150 IZUMO1 GFY LIN7B LOC105447645 RPL18 KASH5 FCGRT PIH1D1 SNAR-A3 PTOV1-AS2 ELSPBP1 SNAR-A4 SNAR-A1 ADM5 SNAR-C2 ODAD1 SAXO3 ZSWIM9 AP2A1 SIGLEC11 SNORD35B BCAT2 PTH2 SNAR-C4 ATF5 KCNA7 MIR4324 SNAR-A2 MIR4749 GRIN2D MED25 SNAR-A11 CARD8-AS1 ZNF473CR HRC TRPM4 GYS1 SNORD33 SNRNP70 FUT1 PTOV1
          nsv833862 Chr.19:49448789 - 49570676 on Build GRCh38 Loss RPL13A NOSIP FCGRT PIH1D1 FLT3LG SNORD35A SNORD32A MIR150 SNORD34 ALDH16A1 SNORD33 RCN3 RPS11 SNORD35B
          nsv833863 Chr.19:49479177 - 49629716 on Build GRCh38 Loss RPL13A NOSIP FCGRT FLT3LG SNORD35A SNORD32A MIR150 SNORD34 PRR12 SNORD33 RCN3 PRRG2 RPS11 SNORD35B
          nsv1160655 Chr.19:49489110 - 49841830 on Build GRCh38 Deletion RPL13A NOSIP FCGRT SNORD35A IRF3 SNORD32A PTOV1-AS1 SNORD34 PRMT1 FUZ SCAF1 PRR12 ADM5 RCN3 RRAS LOC105372435 TSKS AP2A1 SNORD35B MIR5088 MIR6799 BCL2L12 MIR6800 MED25 MIR150 SNORD33 PRRG2 RPS11 CPT1C
          nsv3230349 Chr.19:47957693 - 50112931 on Build GRCh38 Deletion NOSIP DKKL1 GRWD1 PNKP DHDH IRF3 SNORD32A PTOV1-AS1 CYTH2 BAX MIR4750 RRAS CGB2 DBP PPFIA3 MIR5088 MIR6799 BCL2L12 SYNGR4 CD37 SNAR-A14 SNAR-A5 FUT2 PLA2G4C-AS1 SNAR-A8 C19orf73 RPS11 SPHK2 ZNF114 CPT1C RUVBL2 CGB7 SNAR-G2 CGB3 SNAR-A7 LIG1 NTN5 MIR4751 NUCB1-AS1 CA11 AKT1S1 PRMT1 ALDH16A1 FUZ SCAF1 PRR12 RCN3 LOC105372435 TSKS SIGLEC16 TEAD2 LOC105372430 BSPH1 KCNJ14 HSD17B14 SNAR-G1 FGF21 EMP3 PLA2G4C MAMSTR CABP5 SLC17A7 LMTK3 NUP62 LOC101928295 SULT2B1 CGB8 PLEKHA4 TMEM143 IL4I1 PRRG2 RPL13A TULP2 FTL SNAR-A6 FLT3LG SNORD35A MIR6798 FAM83E KDELR1 SNORD34 PPP1R15A ZNF473 NTF4 CGB1 VRK3 RASIP1 SEC1P SPACA4 MIR6800 LHB NUCB1 CGB5 TBC1D17 SLC6A16 CARD8 MIR150 IZUMO1 GFY LIN7B LOC105447645 RPL18 KASH5 FCGRT PIH1D1 PTOV1-AS2 ELSPBP1 SNAR-A4 ADM5 ODAD1 SAXO3 ZSWIM9 AP2A1 SIGLEC11 SNORD35B BCAT2 PTH2 ATF5 KCNA7 MIR4324 MIR4749 GRIN2D MED25 CARD8-AS1 ZNF473CR HRC TRPM4 GYS1 SNORD33 SNRNP70 FUT1 PTOV1

          Back To Top

          More Information


          Set Membership:

          Intragenic DGV Variation

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline