Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04034020_cn
          See other LILRA6 CNV Assays ›
          Gene Symbol
          LILRA6
          Assay Reference Genome
          Location

          Chr.19:54239468 on build GRCh38
          Cytoband
          19q13.42
          Assay ID Hs04034020_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          79168

          Gene Name:

          leukocyte immunoglobulin like receptor A6

          Gene Aliases:

          CD85b, ILT-8, ILT5, ILT8, LILRB3, LILRB6

          Location:

          Chr.19:54236592-54242768 on Build GRCh38

          Assay Gene Location:

          Within Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LILRA6 NM_024318.3 NP_077294.3
          NR_104098.1
          AF041262.1 AAD02204.1
          DA463849.1
          KC918856.1
          KC918859.1
          KC918864.1
          KC918865.1
          KC918866.1
          KC918867.1
          KC918868.1
          KC918869.1
          KC918870.1
          KC918871.1
          KC918872.1
          KC918873.1
          KC918874.1
          KC918875.1
          KC918876.1
          KF294245.1
          KF294247.1
          KF294248.1
          KF294250.1
          KF294251.1
          KF294252.1
          KF294253.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1151876 Chr.19:54238523 - 54246248 on Build GRCh38 Duplication LILRA6
          dgv3654n100 Chr.19:54212959 - 54303818 on Build GRCh38 Gain LILRB5 LILRB2 MIR4752 LILRB3 LILRA6
          nsv9752 Chr.19:54218894 - 54244649 on Build GRCh38 Gain+Loss LILRB3 LILRA6
          esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion MIR512-1 MIR516B2 NLRP12 LENG1 NDUFA3 MIR520G MIR371B MIR520C MIR935 MIR520F ZNF677 MIR515-2 LILRA5 MIR517B MIR518A2 MIR515-1 MIR1323 LILRB5 LILRB2 MIR519B MIR371A MBOAT7 MIR519E MIR526A2 MIR525 MIR518E PRKCG BIRC8 MIR527 MIR1283-2 ZNF845 MIR516B1 ZNF331 MIR520H TPM3P9 TMC4 LILRB3 MIR520D MIR518A1 MYADM MIR518C MIR526B MIR526A1 MIR520B MIR517A MIR523 ZNF765 PRPF31 MIR520A MIR4752 MIR516A2 MIR519A2 MIR1283-1 ZNF525 MIR519D VN1R2 ZNF813 TFPT CACNG8 MIR372 LOC284379 TSEN34 OSCAR MIR516A1 MIR519A1 CNOT3 MIR373 DPRX MIR518B MIR517C LOC105372457 ZNF761 MIR518D RPS9 ZNF665 MIR522 LILRA6 MIR520E ZNF818P MIR521-2 MIR498 MIR519C CACNG6 MIR521-1 FAM90A27P TARM1 CACNG7 MIR512-2 MIR518F MIR524 VSTM1 VN1R4
          dgv1074e212 Chr.19:54218859 - 54292176 on Build GRCh38 Gain LILRB5 LILRB2 MIR4752 LILRB3 LILRA6
          esv2760520 Chr.19:54212971 - 54250521 on Build GRCh38 Gain+Loss LILRB5 LILRB3 LILRA6
          dgv1075e212 Chr.19:54226329 - 54249469 on Build GRCh38 Gain LILRA6
          nsv183 Chr.19:54212629 - 54265267 on Build GRCh38 Deletion LILRB5 LILRB3 LILRA6
          esv3574853 Chr.19:54226365 - 54326343 on Build GRCh38 Gain LILRB5 LILRB2 LILRA5 MIR4752 LILRA6
          esv3644786 Chr.19:54224126 - 54243061 on Build GRCh38 Loss LILRA6
          dgv2026e59 Chr.19:54233964 - 54242586 on Build GRCh38 Duplication LILRA6
          dgv3658n100 Chr.19:54218406 - 54270335 on Build GRCh38 Gain+Loss LILRB5 LILRB3 LILRA6
          esv1006598 Chr.19:54238010 - 54250220 on Build GRCh38 Insertion LILRA6
          nsv1142120 Chr.19:54219911 - 54239664 on Build GRCh38 Deletion LILRB3 LILRA6
          esv2663554 Chr.19:54224745 - 54244579 on Build GRCh38 Deletion LILRA6
          dgv3656n100 Chr.19:54213432 - 54270335 on Build GRCh38 Gain LILRB5 LILRB3 LILRA6
          nsv2548 Chr.19:54239379 - 54261184 on Build GRCh38 Insertion LILRB5 LILRA6
          esv3390177 Chr.19:54224768 - 54243089 on Build GRCh38 Duplication LILRA6
          nsv2546 Chr.19:54182614 - 54270671 on Build GRCh38 Deletion LILRB5 RPS9 MBOAT7 LILRB3 TSEN34 LILRA6
          esv2718860 Chr.19:54223005 - 54312746 on Build GRCh38 Deletion LILRB5 LILRB2 LILRA5 MIR4752 LILRB3 LILRA6
          dgv6551n54 Chr.19:54223279 - 54240481 on Build GRCh38 Loss LILRA6
          dgv77e180 Chr.19:54224577 - 54240655 on Build GRCh38 Gain LILRA6
          dgv1073e212 Chr.19:54218570 - 54249469 on Build GRCh38 Loss LILRB3 LILRA6
          nsv510774 Chr.19:54158011 - 54269511 on Build GRCh38 Deletion LILRB5 TMC4 RPS9 MBOAT7 LILRB3 TSEN34 LILRA6 LENG1
          nsv953615 Chr.19:54238725 - 54256437 on Build GRCh38 Deletion LILRB5 LILRA6
          esv2718859 Chr.19:54219746 - 54239661 on Build GRCh38 Deletion LILRB3 LILRA6
          nsv428371 Chr.19:54163847 - 54366984 on Build GRCh38 Gain+Loss LILRB5 LILRB2 TMC4 RPS9 LILRA4 MBOAT7 LILRA5 MIR4752 LILRB3 TSEN34 LILRA6 LAIR1
          dgv3657n100 Chr.19:54215071 - 54282373 on Build GRCh38 Gain LILRB5 LILRB2 MIR4752 LILRB3 LILRA6
          nsv498906 Chr.19:54223400 - 54244238 on Build GRCh38 Loss LILRA6
          esv1003658 Chr.19:54215069 - 54257872 on Build GRCh38 Gain LILRB5 LILRB3 LILRA6
          nsv978848 Chr.19:54238900 - 54244501 on Build GRCh38 Duplication LILRA6
          dgv598n67 Chr.19:54220589 - 54240454 on Build GRCh38 Gain LILRB3 LILRA6
          nsv513760 Chr.19:54221726 - 54242028 on Build GRCh38 Insertion LILRB3 LILRA6
          dgv6555n54 Chr.19:54225208 - 54243927 on Build GRCh38 Gain LILRA6
          nsv580148 Chr.19:54195523 - 54245159 on Build GRCh38 Loss RPS9 LILRB3 LILRA6
          nsv1115551 Chr.19:54233224 - 54240324 on Build GRCh38 Duplication LILRA6
          dgv6548n54 Chr.19:54219677 - 54243728 on Build GRCh38 Gain LILRB3 LILRA6
          esv23225 Chr.19:54220049 - 54245180 on Build GRCh38 Gain+Loss LILRB3 LILRA6
          esv3893247 Chr.19:54227805 - 54245159 on Build GRCh38 Loss LILRA6

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s)

          adaptive immune response

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline