Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04116576_cn
          See other TMLHE CNV Assays ›
          Gene Symbol
          TMLHE
          Assay Reference Genome
          Location

          Chr.X:155566142 on build GRCh38
          Cytoband
          Xq28
          Assay ID Hs04116576_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55217

          Gene Name:

          trimethyllysine hydroxylase, epsilon

          Gene Aliases:

          AUTSX6, BBOX2, TMLD, TMLH, TMLHED, XAP130

          Location:

          Chr.X:155489007-155612961 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TMLHE NM_001184797.1 NP_001171726.1
          NM_018196.3 NP_060666.1
          XM_017029620.1 XP_016885109.1
          AF373407.1 AAL01871.1
          AK001589.1 BAA91775.1
          AK291694.1
          BC025269.1 AAH25269.1
          DA323339.1
          DB174485.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv510869 Chr.X:155552481 - 155600316 on Build GRCh38 Deletion TMLHE
          nsv953711 Chr.X:155560540 - 155573939 on Build GRCh38 Deletion TMLHE
          nsv519485 Chr.X:155507715 - 155699751 on Build GRCh38 Loss SPRY3 TMLHE
          esv3559520 Chr.X:155560340 - 155573872 on Build GRCh38 Deletion TMLHE
          nsv512705 Chr.X:155560384 - 155574125 on Build GRCh38 Loss TMLHE
          nsv7162 Chr.X:155545842 - 155591711 on Build GRCh38 Deletion TMLHE
          esv3575078 Chr.X:155556315 - 155574027 on Build GRCh38 Loss TMLHE
          nsv820472 Chr.X:155560367 - 155567918 on Build GRCh38 Deletion TMLHE
          esv29972 Chr.X:154781072 - 155699751 on Build GRCh38 Gain SPRY3 MIR1184-2 RAB39B BRCC3 H2AFB2 MIR1184-3 VBP1 TMLHE-AS1 F8 CMC4 F8A2 MTCP1 TMLHE FUNDC2 H2AFB3 F8A1 CLIC2 H2AFB1 MIR1184-1 MPP1 F8A3 SMIM9 LOC101927830
          esv24821 Chr.X:155552998 - 155574053 on Build GRCh38 Gain+Loss TMLHE
          nsv829420 Chr.X:155563604 - 155567654 on Build GRCh38 Loss TMLHE
          nsv515187 Chr.X:155563801 - 155567513 on Build GRCh38 Loss TMLHE
          esv33784 Chr.X:153962665 - 155699588 on Build GRCh38 Gain+Loss SPRY3 ATP6AP1 GDI1 RPL10 FUNDC2 MIR718 CH17-340M24.3 TAZ CLIC2 OPN1LW MPP1 UBL4A FAM223A MIR664B DNASE1L1 MECP2 CTAG1A LOC101927830 BRCC3 OPN1MW3 HCFC1-AS1 G6PD MIR1184-3 MIR3202-2 LOC107987332 SNORA70 SNORA56 GAB3 MIR1184-1 SMIM9 MIR3202-1 IKBKG LOC105373386 H2AFB2 MIR6858 PLXNA3 F8 FLNA HCFC1 SLC10A3 CMC4 F8A2 MTCP1 TMLHE F8A1 SNORA36A IRAK1 TEX28 OPN1MW2 MIR1184-2 RAB39B EMD LAGE3 VBP1 TMEM187 FAM50A FAM3A TMLHE-AS1 OPN1MW TKTL1 H2AFB3 FAM223B H2AFB1 CTAG1B DKC1 CTAG2 F8A3
          nsv520699 Chr.X:155545002 - 155699751 on Build GRCh38 Gain SPRY3 TMLHE
          nsv511652 Chr.X:155551462 - 155586778 on Build GRCh38 Loss TMLHE
          esv2994901 Chr.X:155560339 - 155568339 on Build GRCh38 Deletion TMLHE
          esv1007887 Chr.X:155560007 - 155573754 on Build GRCh38 Deletion TMLHE
          esv2659146 Chr.X:155560528 - 155573651 on Build GRCh38 Deletion TMLHE
          nsv1137701 Chr.X:155560539 - 155569939 on Build GRCh38 Deletion TMLHE

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          hydroxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          carnitine biosynthetic process
          oxidation-reduction process
          iron ion binding
          oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
          L-ascorbic acid binding
          trimethyllysine dioxygenase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline