Search
Search

Entrez Gene ID: | 158801 |
Gene Name: | NFKB activating protein pseudogene 1 |
Gene Aliases: |
CXorf42 |
Location: |
Chr.X:120236452-120245267 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAPP1 | NR_027131.1 | |||
| AK093895.1 | BAC04246.1 | |||
| DB004517.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5887334 | Chr.X:119471365 - 121073821 on Build GRCh38 | Deletion |
|