Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04230740_cn
          See other ACP1 CNV Assays ›
          Gene Symbol
          ACP1
          Assay Reference Genome
          Location

          Chr.2:273603 on build GRCh38
          Cytoband
          Assay ID Hs04230740_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          52

          Gene Name:

          acid phosphatase 1

          Gene Aliases:

          HAAP, LMW-PTP, LMWPTP

          Location:

          Chr.2:264947-278283 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ACP1 NM_004300.4 NP_004291.1
          NM_007099.4 NP_009030.1
          NR_024080.2
          AB209838.1 BAD93075.1
          AK289934.1 BAF82623.1
          AK291861.1 BAF84550.1
          BC007422.2 AAH07422.1
          BC106011.1 AAI06012.1
          BI460080.1
          BJ992018.1
          BT007136.1 AAP35800.1
          DB443811.1
          M87545.1
          S62884.1 AAB27085.1
          Y16846.1 CAA76416.1

          Target Gene Details

          Entrez Gene ID:

          7840

          Gene Name:

          ALMS1 centrosome and basal body associated protein

          Gene Aliases:

          ALSS

          Location:

          Chr.2:55253-279417 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ALMS1

          Target Gene Details

          Entrez Gene ID:

          728378

          Gene Name:

          POTE ankyrin domain family member F

          Gene Aliases:

          A26C1B, ACTB, POTE2alpha, POTEACTIN

          Location:

          Chr.2:241401-305918 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          POTEF

          Target Gene Details

          Entrez Gene ID:

          54884

          Gene Name:

          retinol saturase

          Gene Aliases:

          -

          Location:

          Chr.2:268195-280766 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RETSAT

          Target Gene Details

          Entrez Gene ID:

          6295

          Gene Name:

          S-antigen visual arrestin

          Gene Aliases:

          RP47, RP96, S-AG

          Location:

          Chr.2:253154-292393 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SAG

          Target Gene Details

          Entrez Gene ID:

          8604

          Gene Name:

          solute carrier family 25 member 12

          Gene Aliases:

          AGC1, ARALAR, DEE39, EIEE39

          Location:

          Chr.2:213578-324837 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SLC25A12

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3589598 Chr.2:250064 - 298533 on Build GRCh38 Gain ACP1 ELMOD3 POTEF IGKV2-24 TRIM51JP SLC25A12 ALMS1 SH3YL1 IGK SNORA41 SNORD51 ALKAL2 SAG RETSAT TGOLN2 LOC100506076 IGKV3-20 NDUFS1 CMKLR2 EEF1B2 LOC105373346
          esv3589599 Chr.2:251988 - 283726 on Build GRCh38 Gain ACP1 ELMOD3 POTEF TRIM51JP SLC25A12 ALMS1 SH3YL1 IGK SNORA41 SNORD51 ALKAL2 SAG RETSAT TGOLN2 LOC100506076 NDUFS1 EEF1B2
          dgv3698n100 Chr.2:265970 - 313392 on Build GRCh38 Gain ACP1 ANKRD33BP1 NAT8 ELMOD3 POTEF CMKLR2-AS LINC03052 IGKV2-24 SLC25A12 ALMS1 IGK DGKD SNORA41 SNORD51 ALKAL2 MED15P9 SAG RETSAT LOC100506076 IGKV2-26 NDUFS1 CMKLR2 EEF1B2 LOC105373346
          nsv833159 Chr.2:158491 - 327294 on Build GRCh38 Loss ACP1 FAR2P1 ANKRD36 ALMS1 SH3YL1 ALKAL2 SAG RETSAT SCARNA6 FAHD2A LOC100506076 IGKV2-26 IGKV3-20 NDUFS1 CMKLR2 TPO TRIM43 ALMS1P1 DYNC1I2 ELMOD3 LINC03052 TRIM51JP TCF7L1 LINC01865 IGK LINC01881 SNORA41 INO80D INO80D-AS1 LALTOP ATG16L1 CMKLR2-AS IGKV2-24 SNORD51 MED15P9 TGOLN2 LOC124905368 LOC105373346 GCSHP3 TRIM64FP ANKRD33BP1 NAT8 POTEF SLC25A12 DGKD NDUFA10 SCARNA5 TDRD15 CCDC74B EEF1B2
          nsv4052979 Chr.2:10001 - 590000 on Build GRCh38 Deletion FAR2P1 ANKRD36 TWIST2 CRYGD TRAPPC12-AS1 FAHD2A MIR3606 LOC100506076 IGKV2-26 IGKV3-20 TPO IGKV2-29 LINC01856 LINC03100 TCF7L1-IT1 SNTG2 COL3A1 TCF7L1 LINC01865 IGK NAT8B INO80D-AS1 LINC01822 TUBA3E FAHD2B RAB6C-AS1 LINC02572 GPAT2P2 LOC102724579 ATG16L1 G6PC2 NOC2LP1 SNORD51 ADI1 APPAT MED15P9 TGOLN2 ZDBF2 TPRKB FAM240C LOC124905368 LOC285097 GAL3ST2 NEU4 LOC105373346 TRIM64FP ANKRD33BP1 CRYGC PLAC9P1 GPAT2 SLC25A12 TRAPPC12 SPC25 SCARNA5 NOSTRIN LINC01874 LOC101926959 LOC124905962 EEF1B2 ALMS1-IT1 ACP1 INPP5D RTP5 MIR4784 LINC03124 RAB6C ALMS1 SH3YL1 HAT1 ALKAL2 SAG RETSAT SCARNA6 LOC105373352 NDUFS1 CMKLR2 TRIM43 ALMS1P1 DYNC1I2 METAP1D ELMOD3 C2orf92 LINC03052 TRIM51JP MZT2B ANKRD36B LINC01115 ABCB11 LINC01881 DLX1 SNORA41 INO80D IGKV1-5 C2orf78 LINC01238 LALTOP KCNIP3 PDCD1 COX5B LINC01237 LOC100996637 CMKLR2-AS IGKV2-24 PROM2 LINC01880 TRIM43B DUSP11 SMPD4 CDRT15P3 KIF5C ACTR1B GCSHP3 NAT8 APOB POTEF DGKD LOC100507443 NDUFA10 MIR1245B TDRD15 LOC100287387 LINC01875 MIR1245A CCDC74B FAM110C LOC150935
          nsv6316255 Chr.2:27263 - 377948 on Build GRCh38 Duplication FAR2P1 ANKRD36 TWIST2 CRYGD TRAPPC12-AS1 FAHD2A MIR3606 LOC100506076 IGKV2-26 IGKV3-20 TPO IGKV2-29 LINC01856 LINC03100 TCF7L1-IT1 SNTG2 COL3A1 TCF7L1 LINC01865 IGK NAT8B INO80D-AS1 LINC01822 TUBA3E FAHD2B RAB6C-AS1 LINC02572 GPAT2P2 LOC102724579 ATG16L1 G6PC2 SNORD51 ADI1 MED15P9 TGOLN2 FAM240C LOC124905368 LOC285097 NEU4 LOC105373346 TRIM64FP ANKRD33BP1 CRYGC PLAC9P1 GPAT2 SLC25A12 TRAPPC12 SPC25 SCARNA5 NOSTRIN EEF1B2 ALMS1-IT1 ACP1 INPP5D RTP5 MIR4784 LINC03124 RAB6C ALMS1 SH3YL1 HAT1 ALKAL2 SAG RETSAT SCARNA6 NDUFS1 CMKLR2 TRIM43 ALMS1P1 DYNC1I2 ELMOD3 LINC03052 TRIM51JP MZT2B LINC01115 ABCB11 LINC01881 SNORA41 INO80D IGKV1-5 LALTOP KCNIP3 PDCD1 LINC01237 CMKLR2-AS IGKV2-24 PROM2 LINC01880 TRIM43B SMPD4 CDRT15P3 KIF5C GCSHP3 NAT8 APOB POTEF DGKD LOC100507443 NDUFA10 MIR1245B TDRD15 LOC100287387 MIR1245A CCDC74B FAM110C LOC150935
          nsv6321644 Chr.2:214401 - 293900 on Build GRCh38 Duplication ACP1 ATG16L1 FAR2P1 IGKV2-24 ANKRD36 ALMS1 SH3YL1 SNORD51 ALKAL2 SAG RETSAT SCARNA6 TGOLN2 LOC100506076 IGKV3-20 NDUFS1 CMKLR2 TPO TRIM43 LOC105373346 GCSHP3 ELMOD3 POTEF TRIM51JP SLC25A12 TCF7L1 IGK SNORA41 SCARNA5 EEF1B2
          nsv6321692 Chr.2:264180 - 313687 on Build GRCh38 Duplication ACP1 CMKLR2-AS IGKV2-24 ALMS1 SH3YL1 SNORD51 ALKAL2 MED15P9 SAG RETSAT LOC100506076 IGKV2-26 NDUFS1 CMKLR2 LOC105373346 ANKRD33BP1 NAT8 ELMOD3 POTEF LINC03052 SLC25A12 IGK DGKD SNORA41 EEF1B2
          nsv4059558 Chr.2:56853 - 309597 on Build GRCh38 Duplication ACP1 INPP5D RTP5 LINC03124 FAR2P1 ANKRD36 RAB6C ALMS1 SH3YL1 TWIST2 CRYGD ALKAL2 TRAPPC12-AS1 SAG RETSAT SCARNA6 FAHD2A MIR3606 LOC100506076 IGKV2-26 IGKV3-20 NDUFS1 CMKLR2 TPO TRIM43 LINC01856 LINC03100 TCF7L1-IT1 DYNC1I2 ELMOD3 SNTG2 TRIM51JP COL3A1 TCF7L1 IGK LINC01115 ABCB11 LINC01881 SNORA41 INO80D INO80D-AS1 FAHD2B RAB6C-AS1 LALTOP KCNIP3 PDCD1 GPAT2P2 LOC102724579 LINC01237 ATG16L1 IGKV2-24 G6PC2 LINC01880 SNORD51 ADI1 MED15P9 TGOLN2 CDRT15P3 KIF5C FAM240C LOC124905368 LOC105373346 GCSHP3 CRYGC APOB PLAC9P1 POTEF GPAT2 SLC25A12 TRAPPC12 SPC25 DGKD LOC100507443 NDUFA10 SCARNA5 MIR1245B TDRD15 LOC100287387 MIR1245A LOC150935 EEF1B2 ALMS1-IT1
          esv2759016 Chr.2:167949 - 929902 on Build GRCh38 Gain TMEM18-DT ACP1 MIR4784 FAR2P1 ANKRD36 ALMS1 SH3YL1 HAT1 ALKAL2 SAG TMEM131 RETSAT SCARNA6 FAHD2A LOC100506076 LOC105373352 IGKV2-26 IGKV3-20 NDUFS1 CMKLR2 TPO IGKV2-29 TRIM43 ALMS1P1 DYNC1I2 METAP1D ELMOD3 C2orf92 LINC03052 TRIM51JP MZT2B ANKRD36B TCF7L1 LINC01865 IGK LINC01115 DLX1 NAT8B SNORA41 INO80D INO80D-AS1 ZAP70 TUBA3E C2orf78 LALTOP COX5B LOC100996637 ATG16L1 CMKLR2-AS IGKV2-24 NOC2LP1 LOC105373480 SNORD51 APPAT MED15P9 LINC01939 TRIM43B DUSP11 TGOLN2 SMPD4 ZDBF2 TPRKB ACTR1B LOC124905368 LOC105373346 GCSHP3 TRIM64FP ANKRD33BP1 NAT8 TMEM18 POTEF SLC25A12 DGKD NDUFA10 SCARNA5 TDRD15 LINC01875 CCDC74B LINC01874 LOC101926959 LOC124905962 EEF1B2
          nsv5442514 Chr.2:252077 - 294598 on Build GRCh38 Duplication ACP1 ELMOD3 POTEF IGKV2-24 TRIM51JP SLC25A12 ALMS1 SH3YL1 IGK SNORA41 SNORD51 ALKAL2 SAG RETSAT TGOLN2 LOC100506076 NDUFS1 CMKLR2 EEF1B2 LOC105373346
          dgv3779n223 Chr.2:197016 - 763147 on Build GRCh38 Duplication TMEM18-DT ACP1 MIR4784 FAR2P1 ANKRD36 ALMS1 SH3YL1 HAT1 ALKAL2 SAG TMEM131 RETSAT SCARNA6 LOC100506076 LOC105373352 IGKV2-26 IGKV3-20 NDUFS1 CMKLR2 TPO IGKV2-29 TRIM43 ALMS1P1 METAP1D ELMOD3 C2orf92 LINC03052 TRIM51JP MZT2B ANKRD36B TCF7L1 LINC01865 IGK DLX1 NAT8B SNORA41 ZAP70 TUBA3E C2orf78 COX5B LOC100996637 ATG16L1 CMKLR2-AS IGKV2-24 NOC2LP1 LOC105373480 SNORD51 APPAT MED15P9 TRIM43B DUSP11 TGOLN2 SMPD4 ZDBF2 TPRKB ACTR1B LOC105373346 GCSHP3 TRIM64FP ANKRD33BP1 NAT8 TMEM18 POTEF SLC25A12 DGKD NDUFA10 SCARNA5 TDRD15 LINC01875 CCDC74B LINC01874 LOC101926959 LOC124905962 EEF1B2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          acid phosphatase activity
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          protein binding
          hydrolase activity
          phosphatase activity
          SH3 domain binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline