Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04310590_cn
          See other HLA-DPA1 CNV Assays ›
          Gene Symbol
          HLA-DPA1
          Assay Reference Genome
          Location

          Chr.6:33076573 on build GRCh38
          Cytoband
          6p21.32
          Assay ID Hs04310590_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3113

          Gene Name:

          major histocompatibility complex, class II, DP alpha 1

          Gene Aliases:

          DP(W3), DP(W4), HLA-DP1A, HLADP, HLASB, PLT1

          Location:

          Chr.6:33064569-33080778 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          HLA-DPA1 NM_001242524.1 NP_001229453.1
          NM_001242525.1 NP_001229454.1
          BI553587.1
          CD516536.1

          Target Gene Details

          Entrez Gene ID:

          3115

          Gene Name:

          major histocompatibility complex, class II, DP beta 1

          Gene Aliases:

          DPB1, HLA-DP, HLA-DP1B, HLA-DPB

          Location:

          Chr.6:33075926-33089696 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          HLA-DPB1 NM_002121.5 NP_002112.3
          AB208978.1 BAD92215.1
          AK291986.1
          AK301322.1
          AK303153.1
          AK313305.1
          AY656678.1 AAV71025.1
          AY656679.1 AAV71026.1
          AY656680.1 AAV71027.1
          AY804132.1 AAW78737.1
          AY804133.1 AAW78738.1
          AY804134.1 AAW78739.1
          AY804135.1 AAW78740.1
          AY804136.1 AAW78741.1
          AY804137.1 AAW78742.1
          AY804138.1 AAW78743.1
          AY804139.1 AAW78744.1
          AY804140.1 AAW78745.1
          AY804141.1 AAW78746.1
          AY804142.1 AAW78747.1
          AY804143.1 AAW78748.1
          AY831401.1 AAW80919.1
          AY831402.1 AAW80920.1
          AY831403.1 AAW80921.1
          AY831404.1 AAW80922.1
          BC007963.2 AAH07963.1
          BC013184.1 AAH13184.1
          BC015000.1 AAH15000.1
          BP213121.1
          BT019780.1 AAV38583.1
          DQ284441.1 ABC66207.1
          FR775797.1
          GQ891549.1
          HE603611.1
          HE603612.1
          HE614584.1
          HE614585.1
          HE614586.1
          HE614587.1
          HE614588.1
          HE614589.1
          HE614590.1
          HE614591.1
          J03041.1 AAA35993.1
          KJ780721.1
          M28200.1 AAA53195.1
          M28202.1 AAA53197.1
          M57466.1 AAA63221.1
          M83664.1 AAA59837.1
          X01426.1 CAA25672.1
          X03067.1 CAA26871.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv823645 Chr.6:33061191 - 33091364 on Build GRCh38 Loss HLA-DPB1 HLA-DPA1
          esv2731879 Chr.6:33065144 - 33107309 on Build GRCh38 Deletion HLA-DPB1 HLA-DPA1
          nsv1024429 Chr.6:32997190 - 33154554 on Build GRCh38 Loss HLA-DPB1 HLA-DOA HLA-DPB2 HCG24 HLA-DPA1
          esv1006913 Chr.6:33064893 - 33088398 on Build GRCh38 Gain HLA-DPB1 HLA-DPA1
          nsv1073969 Chr.6:31456422 - 33480523 on Build GRCh38 Deletion DAXX LOC100507547 RGL2 LOC102725068 TNF SKIV2L MSH5 HLA-DPB1 SNORD117 ATP6V1G2 LY6G6F RNF5 BTNL2 HLA-DOB PHF1 COL11A2 C2-AS1 MIR3135B MSH5-SAPCD1 APOM SNORD52 HLA-DQB2 CYP21A1P PPT2 PFDN6 C4B HSPA1A HSD17B8 MICB PPT2-EGFL8 TNXA PSMB9 MIR1236 SLC39A7 HLA-DQA1 C6orf10 ABHD16A RING1 C6orf25 NELFE GPSM3 CFB HLA-DOA ZBTB9 PPP1R2P1 BRD2 LTB HLA-DRA NOTCH4 MIR6873 EHMT2 PRRT1 MIR6834 FKBPL MIR6832 TAP2 RPS18 LY6G6D MIR6721 CYP21A2 ATF6B DDAH2 HLA-DQB1 HSPA1B LY6G6E VWA7 BAG6 C6orf47 WDR46 LOC101929163 EGFL8 MIR4646 HLA-DRB5 CUTA PSMB8 TNXB ATP6V1G2-DDX39B TAP1 VARS CSNK2B HSPA1L GPANK1 TAPBP LY6G5C LSM2 LY6G6C B3GALT4 MIR5004 MCCD1 HLA-DRB6 DXO MIR219A1 NCR3 ZBTB12 SLC44A4 HCG26 LOC100287329 KIFC1 CLIC1 LOC100294145 HCP5 NEU1 C4A SAPCD1 AGER HCG24 STK19 ZBTB22 LY6G5B SNORA38 HLA-DQA2 C2 HLA-DMA PBX2 NFKBIL1 AGPAT1 SAPCD1-AS1 C6orf48 SNORD48 MIR6833 LTA PRRC2A HLA-DQB1-AS1 HLA-DPA1 LST1 SYNGAP1 DDX39B SNORD84 HCG25 PSMB8-AS1 HLA-DRB1 VPS52 HLA-DMB AIF1 DDX39B-AS1 HLA-DPB2 HCG23 RXRB
          nsv1112900 Chr.6:31455323 - 33480423 on Build GRCh38 Deletion DAXX LOC100507547 RGL2 LOC102725068 TNF SKIV2L MSH5 HLA-DPB1 SNORD117 ATP6V1G2 LY6G6F RNF5 BTNL2 HLA-DOB PHF1 COL11A2 C2-AS1 MIR3135B MSH5-SAPCD1 APOM SNORD52 HLA-DQB2 CYP21A1P PPT2 PFDN6 C4B HSPA1A HSD17B8 MICB PPT2-EGFL8 TNXA PSMB9 MIR1236 SLC39A7 HLA-DQA1 C6orf10 ABHD16A RING1 C6orf25 NELFE GPSM3 CFB HLA-DOA ZBTB9 PPP1R2P1 BRD2 LTB HLA-DRA NOTCH4 MIR6873 EHMT2 PRRT1 MIR6834 FKBPL MIR6832 TAP2 RPS18 LY6G6D MIR6721 CYP21A2 ATF6B DDAH2 HLA-DQB1 HSPA1B LY6G6E VWA7 BAG6 C6orf47 WDR46 LOC101929163 EGFL8 MIR4646 HLA-DRB5 CUTA PSMB8 TNXB ATP6V1G2-DDX39B TAP1 VARS CSNK2B HSPA1L GPANK1 TAPBP LY6G5C LSM2 LY6G6C B3GALT4 MIR5004 MCCD1 HLA-DRB6 DXO MIR219A1 NCR3 ZBTB12 SLC44A4 HCG26 LOC100287329 KIFC1 CLIC1 LOC100294145 HCP5 NEU1 C4A SAPCD1 AGER HCG24 STK19 ZBTB22 LY6G5B SNORA38 HLA-DQA2 C2 HLA-DMA PBX2 NFKBIL1 AGPAT1 SAPCD1-AS1 C6orf48 SNORD48 MIR6833 LTA PRRC2A HLA-DQB1-AS1 HLA-DPA1 LST1 SYNGAP1 DDX39B SNORD84 HCG25 PSMB8-AS1 HLA-DRB1 VPS52 HLA-DMB AIF1 DDX39B-AS1 HLA-DPB2 HCG23 RXRB
          esv22467 Chr.6:33055021 - 33087568 on Build GRCh38 Loss HLA-DPB1 HLA-DPA1
          nsv602818 Chr.6:32998165 - 33150041 on Build GRCh38 Loss HLA-DPB1 HLA-DOA HLA-DPB2 HCG24 HLA-DPA1
          nsv1119076 Chr.6:32595823 - 33480623 on Build GRCh38 Deletion RING1 DAXX RGL2 HCG24 CUTA ZBTB22 PSMB8 TAP1 HLA-DQA2 HLA-DPB1 HLA-DOA ZBTB9 PPP1R2P1 BRD2 HLA-DOB HLA-DMA TAPBP MIR6873 B3GALT4 MIR5004 PHF1 COL11A2 MIR6834 MIR3135B MIR219A1 TAP2 HLA-DQB2 RPS18 HLA-DQB1-AS1 HLA-DPA1 PFDN6 SYNGAP1 HLA-DQB1 HCG25 PSMB8-AS1 VPS52 KIFC1 HLA-DMB LOC100294145 HSD17B8 HLA-DPB2 PSMB9 RXRB SLC39A7 WDR46 HLA-DQA1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          positive regulation of interferon-gamma production
          positive regulation of T cell proliferation
          T cell receptor signaling pathway
          positive regulation of T cell activation
          interferon-gamma-mediated signaling pathway
          cellular response to interferon-gamma
          MHC class II receptor activity
          peptide antigen binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline