Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04373227_cn
          See other TCERG1L CNV Assays ›
          Gene Symbol
          TCERG1L
          Assay Reference Genome
          Location

          Chr.10:131113981 on build GRCh38
          Cytoband
          10q26.3
          Assay ID Hs04373227_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          256536

          Gene Name:

          transcription elongation regulator 1 like

          Gene Aliases:

          -

          Location:

          Chr.10:131092391-131311721 on Build GRCh38

          Assay Gene Location:

          Within Intron 11
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TCERG1L NM_174937.3 NP_777597.2
          XM_011539577.1 XP_011537879.1
          AK096269.1
          BC042951.1 AAH42951.1
          BC093639.1 AAH93639.1
          BC101535.1 AAI01536.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3237 Chr.10:131110754 - 131114683 on Build GRCh38 Loss TCERG1L
          nsv832027 Chr.10:131056320 - 131214052 on Build GRCh38 Gain TCERG1L TCERG1L-AS1
          dgv182n67 Chr.10:131110741 - 131114511 on Build GRCh38 Loss TCERG1L
          esv2743528 Chr.10:131110779 - 131114511 on Build GRCh38 Deletion TCERG1L
          esv6315 Chr.10:131110545 - 131114773 on Build GRCh38 Loss TCERG1L
          esv26490 Chr.10:131110462 - 131114883 on Build GRCh38 Loss TCERG1L
          esv32814 Chr.10:131110988 - 131114383 on Build GRCh38 Gain+Loss TCERG1L
          nsv825647 Chr.10:131113787 - 131114368 on Build GRCh38 Loss TCERG1L
          dgv175e199 Chr.10:131110795 - 131114516 on Build GRCh38 Deletion TCERG1L
          dgv1445n54 Chr.10:131112246 - 131114060 on Build GRCh38 Loss TCERG1L
          nsv512193 Chr.10:131110728 - 131114606 on Build GRCh38 Loss TCERG1L
          dgv1443n54 Chr.10:131111671 - 131114439 on Build GRCh38 Loss TCERG1L
          nsv1048253 Chr.10:129847356 - 131188858 on Build GRCh38 Gain GLRX3 LINC00959 EBF3 MIR378C TCERG1L LOC102724883 CTAGE7P TCERG1L-AS1
          esv3547075 Chr.10:131110688 - 131114898 on Build GRCh38 Deletion TCERG1L
          nsv1054934 Chr.10:130334322 - 133158679 on Build GRCh38 Gain CFAP46 NKX6-2 STK32C LINC01164 C10orf91 MIR378C LINC01166 TCERG1L TCERG1L-AS1 LINC01168 ADGRA1-AS1 PWWP2B LINC01167 DPYSL4 LOC105378568 BNIP3 PPP2R2D LOC107984282 LRRC27 ADGRA1 INPP5A JAKMIP3
          nsv1128533 Chr.10:131109637 - 131115537 on Build GRCh38 Deletion TCERG1L
          nsv552442 Chr.10:131112791 - 131114251 on Build GRCh38 Loss TCERG1L
          nsv7621 Chr.10:131081871 - 131117163 on Build GRCh38 Deletion TCERG1L TCERG1L-AS1
          nsv470974 Chr.10:131049527 - 131126683 on Build GRCh38 Gain TCERG1L TCERG1L-AS1
          nsv428242 Chr.10:130607951 - 131583141 on Build GRCh38 Gain MIR378C TCERG1L TCERG1L-AS1
          nsv820144 Chr.10:131110825 - 131114581 on Build GRCh38 Loss TCERG1L
          nsv511463 Chr.10:131110205 - 131116646 on Build GRCh38 Loss TCERG1L
          esv3624952 Chr.10:131110819 - 131114518 on Build GRCh38 Gain TCERG1L

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          general transcription factor

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline