Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04406005_cn
          See other SLC2A3 CNV Assays ›
          Gene Symbol
          SLC2A3
          Assay Reference Genome
          Location

          Chr.12:7928465 on build GRCh38
          Cytoband
          12p13.31
          Assay ID Hs04406005_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6515

          Gene Name:

          solute carrier family 2 member 3

          Gene Aliases:

          GLUT3

          Location:

          Chr.12:7919228-7936296 on Build GRCh38

          Assay Gene Location:

          Within Intron 6
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SLC2A3 NM_006931.2 NP_008862.1
          AB209607.1 BAD92844.1
          AK298673.1
          AK304509.1
          AK312386.1
          BC039196.1 AAH39196.1
          CR457413.1 CAG33694.1
          M20681.1 AAB61083.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv2334n54 Chr.12:7847740 - 7950607 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          esv2751114 Chr.12:7810616 - 7964473 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          esv3580097 Chr.12:7851816 - 7971452 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          nsv826225 Chr.12:7845109 - 7974566 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv59e55 Chr.12:7864877 - 7989987 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          esv29743 Chr.12:7842682 - 7970710 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv61n111 Chr.12:7811433 - 7978362 on Build GRCh38 Duplication SLC2A3 NANOGP1 SLC2A14
          esv3628450 Chr.12:7916707 - 7930712 on Build GRCh38 Gain SLC2A3
          dgv2338n54 Chr.12:7900980 - 7970710 on Build GRCh38 Loss SLC2A3
          nsv469100 Chr.12:7871766 - 7946706 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          nsv470265 Chr.12:7840720 - 7970710 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          nsv470264 Chr.12:7832345 - 7973148 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv2335n54 Chr.12:7848316 - 7973149 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          nsv557331 Chr.12:7923089 - 7970710 on Build GRCh38 Gain SLC2A3
          esv3548706 Chr.12:7842215 - 7980904 on Build GRCh38 Deletion SLC2A3 NANOGP1 SLC2A14
          esv3580549 Chr.12:7918219 - 8055080 on Build GRCh38 Gain SLC2A3 FOXJ2
          dgv2337n54 Chr.12:7888325 - 7973149 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          esv3628445 Chr.12:7847526 - 8064130 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14 C3AR1 FOXJ2
          dgv58e55 Chr.12:7801455 - 7987805 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv2336n54 Chr.12:7869336 - 7938243 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          esv2759877 Chr.12:7704223 - 8471391 on Build GRCh38 Gain+Loss SLC2A3 FAM90A1 NECAP1 FAM86FP CLEC4C NANOG LOC101927905 LINC00937 NANOGNB CLEC4A C3AR1 ZNF705A DPPA3 FOXJ2 NANOGP1 FAM66C POU5F1P3 SLC2A14 CLEC6A
          esv3892132 Chr.12:7884840 - 7949790 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv2329n54 Chr.12:7808980 - 7993566 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          esv3628448 Chr.12:7880733 - 7960581 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          dgv60e55 Chr.12:7864882 - 7982073 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          dgv11e213 Chr.12:7851829 - 7987805 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv1354n100 Chr.12:7842179 - 7952475 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv166e201 Chr.12:7827227 - 7939801 on Build GRCh38 Deletion SLC2A3 NANOGP1 SLC2A14
          dgv179n27 Chr.12:7818125 - 7946706 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv2328n54 Chr.12:7800493 - 7935631 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          esv2760250 Chr.12:7842191 - 7978362 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv167e201 Chr.12:7842493 - 7972788 on Build GRCh38 Deletion SLC2A3 NANOGP1 SLC2A14
          esv3548709 Chr.12:7882905 - 7957904 on Build GRCh38 Deletion SLC2A3 NANOGP1 SLC2A14
          dgv449e212 Chr.12:7821394 - 7989987 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv1352n100 Chr.12:7805181 - 7978362 on Build GRCh38 Gain SLC2A3 NANOGP1 SLC2A14
          dgv183n27 Chr.12:7844294 - 7973149 on Build GRCh38 Loss SLC2A3 NANOGP1 SLC2A14
          dgv57e55 Chr.12:7700916 - 7963472 on Build GRCh38 Gain SLC2A3 NANOGP1 CLEC4C SLC2A14 NANOG NANOGNB DPPA3
          dgv5e196 Chr.12:7621924 - 7954221 on Build GRCh38 Duplication SLC2A3 NANOGP1 APOBEC1 CLEC4C SLC2A14 NANOG NANOGNB DPPA3 GDF3

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glucose transport
          L-ascorbic acid metabolic process
          dehydroascorbic acid transport
          glucose transmembrane transport
          glucose transmembrane transporter activity
          glucose binding
          dehydroascorbic acid transporter activity
          D-glucose transmembrane transporter activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline