Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04487500_cn
          See other CYP4F24P CNV Assays ›
          Gene Symbol
          CYP4F24P
          Assay Reference Genome
          Location

          Chr.19:15763201 on build GRCh38
          Cytoband
          Assay ID Hs04487500_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          388514

          Gene Name:

          cytochrome P450 family 4 subfamily F member 24, pseudogene

          Gene Aliases:

          -

          Location:

          Chr.19:15759334-15779988 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CYP4F24P NR_033864.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1060711 Chr.19:15745103 - 15817596 on Build GRCh38 Gain CYP4F24P OR10H5 OR10H1
          nsv6534232 Chr.19:15740705 - 15815936 on Build GRCh38 Duplication CYP4F24P OR10H3 OR10H5 OR10H1
          nsv4740645 Chr.19:15707370 - 15907509 on Build GRCh38 Deletion CYP4F24P CLEC4OP LOC124900418 UCA1 OR10H3 OR10H5 OR10H2 OR10H1 CYP4F2
          esv3334671 Chr.19:15673766 - 15931354 on Build GRCh38 Duplication CYP4F12 CYP4F24P CLEC4OP CYP4F11 LOC124900418 UCA1 OR10H3 OR10H5 OR10H2 OR10H1 CYP4F2
          nsv5939642 Chr.19:15763126 - 15763179 on Build GRCh38 Deletion CYP4F24P
          nsv3248463 Chr.19:15703465 - 15794126 on Build GRCh38 Insertion CYP4F24P OR10H3 OR10H5 OR10H2
          nsv4259346 Chr.19:15654672 - 15801387 on Build GRCh38 Deletion CYP4F12 CYP4F24P OR10H3 OR10H5 CYP4F3 OR10H2
          nsv833766 Chr.19:15759095 - 15956904 on Build GRCh38 Loss CYP4F24P CLEC4OP CYP4F11 LOC124900418 UCA1 OR10H4 OR10H5 OR10H1 CYP4F2
          nsv1064684 Chr.19:15311319 - 16062011 on Build GRCh38 Gain LINC00905 CLEC4OP CYP4F11 BRD4 AKAP8L LOC124900418 UCA1 AKAP8 CYP4F3 LINC00661 OR10H2 OR10H1 RASAL3 PGLYRP2 CYP4F12 WIZ CYP4F24P CYP4F22 MIR1470 CYP4F8 OR10H4 OR10H3 OR10H5 CYP4F2
          esv2718244 Chr.19:15743561 - 15951200 on Build GRCh38 Deletion CYP4F24P CLEC4OP CYP4F11 LOC124900418 UCA1 OR10H4 OR10H5 OR10H1 CYP4F2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline