Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04544840_cn
          See other PPOX CNV Assays ›
          Gene Symbol
          PPOX
          Assay Reference Genome
          Location

          Chr.1:161167016 on build GRCh38
          Cytoband
          Assay ID Hs04544840_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5498

          Gene Name:

          protoporphyrinogen oxidase

          Gene Aliases:

          PPO, V290M, VP, VPCO

          Location:

          Chr.1:161165728-161178013 on Build GRCh38

          Assay Gene Location:

          Within Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PPOX NM_000309.5 NP_000300.1
          NM_001122764.3 NP_001116236.1
          NM_001350128.2 NP_001337057.1
          NM_001350129.2 NP_001337058.1
          NM_001350130.2 NP_001337059.1
          NM_001350131.2 NP_001337060.1
          NM_001365398.1 NP_001352327.1
          NM_001365399.1 NP_001352328.1
          NM_001365400.1 NP_001352329.1
          NM_001365401.1 NP_001352330.1
          XM_005245291.5 XP_005245348.2
          XM_006711404.5 XP_006711467.3
          XM_011509663.3 XP_011507965.1
          XM_011509664.2 XP_011507966.1
          XM_011509665.3 XP_011507967.1
          XM_011509667.3 XP_011507969.1
          XM_011509668.3 XP_011507970.1
          XM_011509670.3 XP_011507972.1
          XM_011509671.2 XP_011507973.1
          XM_011509672.4 XP_011507974.2
          XM_011509673.3 XP_011507975.1
          XM_011509674.3 XP_011507976.1
          XM_024447864.2 XP_024303632.2
          XM_047423569.1 XP_047279525.1
          XM_047423581.1 XP_047279537.1
          AK094855.1 BAG52942.1
          AK298912.1 BAG61019.1
          AK302298.1 BAG63638.1
          BC002357.2 AAH02357.1
          BX384814.2
          D38537.1 BAA07538.1
          DA268550.1
          DA745403.1
          DC329287.1
          DC399187.1
          EL954312.1
          U26446.1 AAA67690.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6332269 Chr.1:160388767 - 162811116 on Build GRCh38 Duplication NOS1AP DEDD SH2D1B TSTD1 LY9 LOC101928372 UAP1 ITLN1 MIR4654 HSD17B7 NIT1 FCRLB LOC100422526 C1orf226 ATF6-DT SPATA46 USP21 HSPA7 DDR2 CFAP126 ITLN2 UFC1 CD84 MIR5187 OLFML2B CD244 FCGR3A SLAMF7 ADAMTS4 FCGR2B FCGR3B SLAMF1 UHMK1 NR1I3 F11R HSPA6 MPZ NECTIN4-AS1 CD48 KLHDC9 NECTIN4 APOA2 TOMM40L MIR556 SLAMF6 LOC105371470 FCER1G LOC112543491 PFDN2 B4GALT3 PPOX FCGR2C DUSP12 CD84-AS1 NDUFS2 PCP4L1 LOC124904439 ATF6 RPL31P11 USF1 ARHGAP30 VANGL2 SDHC FCGR2A FCRLA
          nsv4518029 Chr.1:160848909 - 161416210 on Build GRCh38 Duplication DEDD TSTD1 MPZ NECTIN4-AS1 LOC101928372 KLHDC9 NECTIN4 APOA2 ITLN1 TOMM40L NIT1 FCER1G LOC112543491 PFDN2 B4GALT3 USP21 PPOX CFAP126 ITLN2 UFC1 NDUFS2 PCP4L1 MIR5187 CD244 ADAMTS4 USF1 ARHGAP30 SDHC NR1I3 F11R

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          porphyrin-containing compound biosynthetic process
          protoporphyrinogen IX biosynthetic process
          heme biosynthetic process
          heme a biosynthetic process
          heme B biosynthetic process
          response to xenobiotic stimulus
          protoporphyrinogen IX metabolic process
          heme O biosynthetic process
          oxygen-dependent protoporphyrinogen oxidase activity
          oxidoreductase activity
          flavin adenine dinucleotide binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline