Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04678816_cn
          See other KRCC1 CNV Assays ›
          Gene Symbol
          KRCC1
          Assay Reference Genome
          Location

          Chr.2:88040648 on build GRCh38
          Cytoband
          Assay ID Hs04678816_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          51315

          Gene Name:

          lysine rich coiled-coil 1

          Gene Aliases:

          CHBP2

          Location:

          Chr.2:88027205-88055783 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          KRCC1 NM_001304526.2 NP_001291455.1
          NM_016618.3 NP_057702.1
          XM_017004292.3 XP_016859781.1
          AF208845.1 AAF64259.1
          AF217520.1 AAF67631.1
          BC015927.1 AAH15927.1
          BC107580.1 AAI07581.1
          DB461504.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv61n16 Chr.2:87343737 - 90254660 on Build GRCh38 Deletion IGKV2-29 EIF2AK3-DT IGKV1D-8 LOC124907854 IGKV1-5 IGKV2-24 IGKV2-26 IGKV3D-15 RNU2-63P MALLP2 ANAPC1P4 SMYD1 ANKRD36BP2 MIR4436A IGKV3-20 SPMIP9 FOXI3 LOC107985911 FABP1 MIR4435-1 RPIA EIF2AK3 KMT2CP4 PLGLB2 RGPD2 IGK CYTOR LINC01943 IGKV1D-13 KRCC1 IGKC MIR4780 EIF2AK3-AS1 NCAL1 THNSL2
          nsv4519735 Chr.2:87991480 - 88311481 on Build GRCh38 Duplication RNU2-63P FABP1 SMYD1 KRCC1 MIR4780 THNSL2
          nsv955227 Chr.2:88022682 - 88054981 on Build GRCh38 Duplication KRCC1
          nsv1072005 Chr.2:87360367 - 90256528 on Build GRCh38 Deletion IGKV2-29 EIF2AK3-DT IGKV1D-8 IGKV1-5 IGKV2-24 IGKV2-26 IGKV3D-15 RNU2-63P MALLP2 ANAPC1P4 SMYD1 ANKRD36BP2 MIR4436A IGKV3-20 SPMIP9 FOXI3 LOC107985911 FABP1 MIR4435-1 RPIA EIF2AK3 KMT2CP4 PLGLB2 RGPD2 IGK CYTOR LINC01943 IGKV1D-13 KRCC1 IGKC MIR4780 EIF2AK3-AS1 NCAL1 THNSL2
          dgv6947n54 Chr.2:87815738 - 88850700 on Build GRCh38 Gain FABP1 EIF2AK3-DT RPIA EIF2AK3 RNU2-63P MALLP2 SMYD1 RGPD2 ANKRD36BP2 MIR4436A KRCC1 SPMIP9 FOXI3 MIR4780 EIF2AK3-AS1 THNSL2
          nsv1148611 Chr.2:87360364 - 90256525 on Build GRCh38 Deletion IGKV2-29 EIF2AK3-DT IGKV1D-8 IGKV1-5 IGKV2-24 IGKV2-26 IGKV3D-15 RNU2-63P MALLP2 ANAPC1P4 SMYD1 ANKRD36BP2 MIR4436A IGKV3-20 SPMIP9 FOXI3 LOC107985911 FABP1 MIR4435-1 RPIA EIF2AK3 KMT2CP4 PLGLB2 RGPD2 IGK CYTOR LINC01943 IGKV1D-13 KRCC1 IGKC MIR4780 EIF2AK3-AS1 NCAL1 THNSL2
          nsv582384 Chr.2:87987149 - 88773214 on Build GRCh38 Loss FABP1 EIF2AK3-DT RPIA EIF2AK3 RNU2-63P SMYD1 RGPD2 ANKRD36BP2 KRCC1 SPMIP9 FOXI3 MIR4780 EIF2AK3-AS1 THNSL2
          nsv582383 Chr.2:87915816 - 88527080 on Build GRCh38 Gain RNU2-63P FABP1 SMYD1 RGPD2 KRCC1 SPMIP9 FOXI3 MIR4780 THNSL2
          nsv582386 Chr.2:88035188 - 89208087 on Build GRCh38 Loss FABP1 EIF2AK3-DT RPIA EIF2AK3 IGKV1-5 IGKV2-24 IGKV2-26 MALLP2 SMYD1 IGK ANKRD36BP2 MIR4436A IGKV3-20 KRCC1 IGKC SPMIP9 FOXI3 MIR4780 EIF2AK3-AS1 THNSL2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline