Search
Search

Entrez Gene ID: | 10063 |
Gene Name: | cytochrome c oxidase copper chaperone COX17 |
Gene Aliases: |
- |
Location: |
Chr.3:119663975-119677406 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX17 | NM_001382002.1 | NP_001368931.1 | ||
| NM_005694.2 | NP_005685.1 | |||
| NR_167771.1 | ||||
| NR_167772.1 | ||||
| BC010933.1 | AAH10933.1 | |||
| BC105280.1 | AAI05281.1 | |||
| L77701.1 | AAA98114.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014394 | Chr.3:119641659 - 119693428 on Build GRCh38 | Loss |
|
| nsv6368772 | Chr.3:119640731 - 119691505 on Build GRCh38 | Deletion |
|
| esv3568977 | Chr.3:119657752 - 119694492 on Build GRCh38 | Loss |
|
| nsv591390 | Chr.3:119665316 - 119740004 on Build GRCh38 | Gain |
|
| nsv5889494 | Chr.3:119662960 - 119757421 on Build GRCh38 | Duplication |
|
| nsv6371858 | Chr.3:119672301 - 119679200 on Build GRCh38 | Duplication |
|
| nsv6357308 | Chr.3:119617774 - 119678928 on Build GRCh38 | Deletion |
|
Set Membership: |
|